Search
Search

Tmem91| Gene Symbol: Tmem91 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290497.1 | NP_001277426.1 | 2 - 3 | 368 | 96 | |
| NM_001290498.1 | NP_001277427.1 | 2 - 3 | 368 | 96 | ||
| NM_177102.4 | NP_796076.1 | 2 - 3 | 368 | 96 | ||
| GenBank mRNA | AK043858.1 | - | 2 - 3 | 321 | 96 | |
| AK079312.1 | - | 2 - 3 | 369 | 96 | ||
| BC058996.1 | - | 2 - 3 | 317 | 5711063 | 96 | |
| BE863748.1 | - | 1 - 2 | 249 | 96 | ||
| BQ555196.1 | - | 2 - 3 | 296 | 96 | ||
Set Membership: |
|