Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00622790_m1
          See other GM16897 GE Assays ›
          Gene Symbol:
          Gm16897
          Gene Name:
          predicted gene, 16897
          Gene Aliases:
          C030002c11rik
          Chromosome Location:
          Chr.1: 194955759 - 194976959 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00622790_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gm16897
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_033564.1 2 - 3 1025 114
          GenBank mRNA AK030377.1 - 2 - 3 1026 114

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0100003850.mm.1

          Set Membership:

          Amplicon length greater than or equal to 101 Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 464061 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 847139 BC066020.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 847139 NM_001005788.2 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 156731 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 167466 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 729336 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 953288 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 97119 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 155492 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 137099 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P143371 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P00290472 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P143371 - -
          Whole Mouse Genome G4122A A_52_P471340 AK030377.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P471340 NR_033564.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P143371 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00201121 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20958843 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20958839 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20958807 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20958825 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00640471 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20958809 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20958788 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20958829 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00201138 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20958826 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20958820 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20958817 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00201139 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20958816 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20958822 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20958800 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20958805 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00201117 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20958803 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00230307 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00201128 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20958834 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20958811 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20958837 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20958828 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00294274 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20958833 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00201140 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20958842 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0100031509.mm.1 NR_033564.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100031509.mm.1 NR_033564.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100031509.mm.1 TC0100003850.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 877583 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline