Search
Search

Atp8b5| Gene Symbol: Atp8b5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177195.3 | NP_796169.2 | 6 - 7 | 466 | 85 | |
| GenBank mRNA | AK029992.1 | - | 7 - 8 | 696 | 85 | |
| AK076708.1 | - | 6 - 7 | 467 | 85 | ||
| AK078567.1 | - | 6 - 7 | 435 | 85 | ||
| EF377342.1 | - | 6 - 7 | 455 | 85 | ||
Set Membership: |
|