Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00623514_s1
          See other 9530056K15RIK GE Assays ›
          Gene Symbol:
          9530056K15Rik
          Gene Name:
          RIKEN cDNA 9530056K15 gene
          Gene Aliases:
          -
          Chromosome Location:
          -
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00623514_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 9530056K15Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK035496.1 - 1740 126
          AK035765.1 - 126

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Recommended Assay Mus musculus 1700 Inventoried

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24156420 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24156417 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24156418 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24156414 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24156419 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2360271 AK035765.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2360271 AI854452.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2360271 AK035496.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P109554 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P115307 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24156407 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24156402 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24156399 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24156414 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24156404 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24156412 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24156408 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24156409 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P115307 - -
          Whole Mouse Genome G4122A A_52_P75046 AK035765.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P75046 AK035496.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P121148 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P115307 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24156419 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24156418 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24156417 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24156414 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24156420 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10535468 - -
          Affymetrix Mouse Gene 1.0 ST Array 10535469 - -
          Affymetrix Mouse Gene 1.0 ST Array 10535470 - -
          Affymetrix HT MG-430 PM Array Plate 1457523_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10535467 - -
          Mouse Genome 430 2.0 Array 1457523_at - -
          Mouse Genome 430 2.0 Array 1457523_PM_at - -
          Mouse Expression Set 430 1457523_at - -
          Mouse Expression Set 430 1457523_PM_at - -
          Murine Genome U74v2 Set 136691_PM_at - -
          Murine Genome U74v2 Set 136691_at - -
          Affymetrix HT Mouse Genome 430B Array 1457523_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1457523_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 358687 - -
          ABI Mouse Genome Survey Microarray 723911 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 102850215 - -
          MouseWG-6 v2 Expression BeadChip 6770164 - -
          Illumina mouse-6 v1.0 expression beadchip 6770164 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006770164 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1260155 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1260155 - -
          mouse-6 v1.1 (Illumina) scl25804.1.1_46-S - -
          Illumina Sentrix mouse-6.v1 bead array 6770164 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl25804.1.1_46-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6770164 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1260155 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline