Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Mm00624342_m1
See other ABCA13 GE Assays ›
Gene Symbol:
Abca13
Gene Name:
ATP-binding cassette, sub-family A (ABC1), member 13
Gene Aliases:
9830132L24, A930002G16Rik, AI956815
Chromosome Location:
Chr.11: 9191354 - 9684871 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID
Mm00624342_m1
Best Coverage
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 250 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Inventoried
Catalog #
4331182
Price
284.00
Your Price
Online offer:
266.65
266.65
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: Abca13
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_178259.3
NP_839990.2
1 - 2
91
73
GenBank mRNA
AK036546.1
-
1 - 2
92
73
Back To Top
More Information
Updated Mapping Information
This assay targets the same location as MTA transcript cluster(s) TC1100000120.mm.1
Set Membership:
Ambion siRNA available
Amplicon length between 71 and 85
Transporter
Recommended Assay
Mus musculus
1700
Inventoried
Probe spans exons
Matching Ambion siRNAs:
Click here to find siRNAs for Gene Abca13
Panther Classification:
Molecular Function -
ATP-binding cassette (ABC) transporter
primary active transporter
transporter
Gene Ontology Categories:
Function(s)
Process(es)
transport
biological_process
cholesterol efflux
phospholipid efflux
transmembrane transport
nucleotide binding
transporter activity
ATP binding
phospholipid transporter activity
ATPase activity
cholesterol transporter activity
ATPase activity, coupled to transmembrane movement of substances
MicroArray Probes:
Affymetrix
ABI
Array Name
Probe IDs
Targets
Target Type
Affymetrix Mouse Clariom S Array
PSR1100001214.mm.1
NM_178259.3.1
Refseq
Affymetrix Mouse Clariom S Array
PSR1100001214.mm.1
NM_178259.3
Refseq
Affymetrix Mouse Clariom S Array
PSR1100001214.mm.1
TC1100000120.mm.1
MTA TC
Affymetrix Mouse Clariom S Array
PSR1100001212.mm.1
NM_178259.3.1
Refseq
Affymetrix Mouse Clariom S Array
PSR1100001212.mm.1
NM_178259.3
Refseq
Affymetrix Mouse Clariom S Array
PSR1100001212.mm.1
TC1100000120.mm.1
MTA TC
Array Name
Probe IDs
Targets
Target Type
ABI Mouse Genome Survey Microarray
476340
-
-
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-7d94cb4b65-6wvps:80/100.66.75.98:80.
git-commit: c9e08c96761173abe34e68f880379696776a4827
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.44.1-2026.02.97.1.0