Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00624579_m1
          See other CLEC12A GE Assays ›
          Gene Symbol:
          Clec12a
          Gene Name:
          C-type lectin domain family 12, member a
          Gene Aliases:
          CLL-1, D230024O04, KLRL1, Micl
          Chromosome Location:
          Chr.6: 129349779 - 129365303 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00624579_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Clec12a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_177686.4 NP_808354.1 1 - 2 271 103
          GenBank mRNA AK084335.1 - 1 - 2 229 103
          BC094904.1 - 1 - 2 246 30044572 103

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0600001532.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Clec12a

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of immune response
          protein phosphatase binding
          carbohydrate binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P66520 NM_177686.4 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P66520 AK084335.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P66520 NM_177686.4 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P66520 AK084335.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 521562 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 49655 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 579361 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 142703 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 640886 AB045293.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 640886 AK044384.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 640886 AK079893.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 640886 BC005722.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 640886 NM_021793.1 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 537462 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 885252 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 87253 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 124714 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P157242 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P122556 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P66520 NM_177686.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P66520 AK084335.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P66520 NM_177686.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P66520 AK084335.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24842965 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24842983 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24842967 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24842984 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24842968 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24842977 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24842985 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24842973 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24842963 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24842970 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24842966 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24842980 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24842987 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24842971 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24842982 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24842976 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P157242 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06067735 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P122556 - -
          Whole Mouse Genome G4122A A_52_P66520 NM_177686.4 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P66520 AK084335.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P66520 NM_177686.4 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P66520 AK084335.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P137354 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P122556 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P157242 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05469574 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05775692 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05290089 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24843006 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24843051 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24842966 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24842976 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24843058 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24843014 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24843054 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24842998 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24843066 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24843073 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24843030 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24842948 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24843068 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24842980 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24843047 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24843045 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24843012 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24843031 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24843063 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24842995 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24842972 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24842952 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24842988 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24843076 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24843007 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24843059 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24843050 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24843040 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24842951 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24843064 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24843005 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24843099 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24843025 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24843002 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24842987 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24843071 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24843093 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24843065 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24843090 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24843108 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24842977 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24842963 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24842943 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24843101 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24843038 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24843103 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24843098 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24842984 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24842968 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24843018 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24843061 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24842950 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24843044 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24842970 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24842990 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24842971 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24843067 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24843052 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24843020 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24842991 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24843023 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24842967 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24842982 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24842993 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24843003 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24843049 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24843078 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24843106 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24842945 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24843104 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24843056 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24843000 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24842953 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24842941 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24843082 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24842968 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24842998 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24842950 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24842948 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24842980 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05697426 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24842957 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24842990 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24842955 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24842970 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05812082 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24843000 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24843020 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24843029 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24842982 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24843031 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24842977 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24843037 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05841790 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24843034 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24843005 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05572932 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24842958 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24842953 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24843002 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05053882 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24843010 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05854213 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24842967 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24842971 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05290088 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24843016 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24843018 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24843017 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05182416 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24843011 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24842963 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24843003 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24842983 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24842987 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24842988 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24842993 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24843012 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24842991 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24842984 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24842951 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24842972 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24842995 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24842952 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10542170 - -
          Affymetrix Mouse Gene 1.0 ST Array 10542168 - -
          Affymetrix Mouse Gene 1.0 ST Array 10542165 - -
          Affymetrix Mouse Gene 1.0 ST Array 10542166 - -
          Affymetrix Mouse Gene 1.0 ST Array 10542167 - -
          Affymetrix Mouse Gene 1.0 ST Array 10542171 - -
          Affymetrix Mouse Gene 1.0 ST Array 10542169 - -
          Affymetrix Mouse Gene 1.1 ST Array 10542164 - -
          Affymetrix Mouse Clariom S Array PSR0600012474.mm.1 NM_177686.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600012474.mm.1 NM_177686.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0600012474.mm.1 TC0600001532.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600012472.mm.1 NM_177686.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600012472.mm.1 NM_177686.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0600012472.mm.1 TC0600001532.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600012473.mm.1 NM_177686.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600012473.mm.1 NM_177686.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0600012473.mm.1 TC0600001532.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600012475.mm.1 NM_177686.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600012475.mm.1 NM_177686.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0600012475.mm.1 TC0600001532.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600012477.mm.1 NM_177686.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600012477.mm.1 NM_177686.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0600012477.mm.1 TC0600001532.mm.1 MTA TC
          Murine Genome U74v2 Set 135501_at - -
          Murine Genome U74v2 Set 135501_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 704890 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 2370619 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002370619 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1230533 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1230533 - -
          mouse-6 v1.1 (Illumina) scl29466.8.1_228-S - -
          Illumina Sentrix mouse-6.v1 bead array 2370619 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl29466.8.1_228-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2370619 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1230533 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1230533 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline