| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Gene 1.0 ST Array |
10538113 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10538110 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10538111 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10538114 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10538112 |
- |
- |
| Affymetrix HT MG-430 PM Array Plate |
1442583_PM_a_at |
- |
- |
| Affymetrix HT MG-430 PM Array Plate |
1435048_PM_at |
- |
- |
| Affymetrix HT MG-430 PM Array Plate |
1440724_PM_at |
- |
- |
| Affymetrix HT MG-430 PM Array Plate |
1435049_PM_s_at |
- |
- |
| Affymetrix Mouse Gene 1.1 ST Array |
10538109 |
- |
- |
| Mouse Genome 430 2.0 Array |
1435048_at |
- |
- |
| Mouse Genome 430 2.0 Array |
1435049_s_at |
- |
- |
| Mouse Genome 430 2.0 Array |
1440724_at |
- |
- |
| Mouse Genome 430 2.0 Array |
1442583_a_at |
- |
- |
| Mouse Genome 430 2.0 Array |
1440724_PM_at |
- |
- |
| Mouse Genome 430 2.0 Array |
1442583_PM_a_at |
- |
- |
| Mouse Genome 430 2.0 Array |
1435049_PM_s_at |
- |
- |
| Mouse Genome 430 2.0 Array |
1435048_PM_at |
- |
- |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array |
243373_at |
- |
- |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array |
243373_PM_at |
- |
- |
| Affymetrix Mouse Clariom S Array |
PSR0600004310.mm.1 |
NR_027236.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600004310.mm.1 |
NR_027236.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600004310.mm.1 |
TC0600000563.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600004309.mm.1 |
NR_027236.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600004309.mm.1 |
NR_027236.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600004309.mm.1 |
TC0600000563.mm.1 |
MTA TC |
| Mouse Expression Set 430 |
1440724_PM_at |
- |
- |
| Mouse Expression Set 430 |
1442583_PM_a_at |
- |
- |
| Mouse Expression Set 430 |
1435048_PM_at |
- |
- |
| Mouse Expression Set 430 |
1435049_PM_s_at |
- |
- |
| Mouse Expression Set 430 |
1440724_at |
- |
- |
| Mouse Expression Set 430 |
1435049_s_at |
- |
- |
| Mouse Expression Set 430 |
1435048_at |
- |
- |
| Mouse Expression Set 430 |
1442583_a_at |
- |
- |
| Murine Genome U74v2 Set |
132602_at |
- |
- |
| Murine Genome U74v2 Set |
138936_PM_s_at |
- |
- |
| Murine Genome U74v2 Set |
132602_PM_at |
- |
- |
| Murine Genome U74v2 Set |
138936_s_at |
- |
- |
| Murine Genome U74v2 Set |
138937_at |
- |
- |
| Murine Genome U74v2 Set |
138937_PM_at |
- |
- |
| Affymetrix HT Mouse Genome 430B Array |
1440724_at |
- |
- |
| Affymetrix HT Mouse Genome 430B Array |
1435048_PM_at |
- |
- |
| Affymetrix HT Mouse Genome 430B Array |
1435048_at |
- |
- |
| Affymetrix HT Mouse Genome 430B Array |
1435049_s_at |
- |
- |
| Affymetrix HT Mouse Genome 430B Array |
1440724_PM_at |
- |
- |
| Affymetrix HT Mouse Genome 430B Array |
1435049_PM_s_at |
- |
- |
| Affymetrix HT Mouse Genome 430B Array |
1442583_a_at |
- |
- |
| Affymetrix HT Mouse Genome 430B Array |
1442583_PM_a_at |
- |
- |