Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00625095_m1
          See other WDR66 GE Assays ›
          Gene Symbol:
          Wdr66
          Gene Name:
          WD repeat domain 66
          Gene Aliases:
          4930415N18Rik, 4933428F06Rik, 5031404N07
          Chromosome Location:
          Chr.5: 123254560 - 123327484 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00625095_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Wdr66
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK030281.1 - 5 - 6 2693 82
          BC138176.1 - 5 - 6 825 8861195 82

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0500001412.mm.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2082096 AK016965.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P517174 BC138176.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P517174 AK030281.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P517174 BC138176.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P517174 AK030281.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P147072 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P170719 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2082096 AK016965.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P517174 BC138176.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P517174 AK030281.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P517174 BC138176.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P517174 AK030281.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P147072 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P170719 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05726898 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05726861 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05726830 - -
          Whole Mouse Genome G4122A A_52_P536768 AK015152.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P641267 AK016965.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P517174 BC138176.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P517174 AK030281.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P517174 BC138176.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P517174 AK030281.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P119269 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P147072 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P170719 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24054777 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04223568 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24054896 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24054932 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24054950 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24054894 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04536260 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04591616 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24054824 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24054978 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24054887 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24054940 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24054984 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04223564 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24054989 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04173698 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04710385 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24054809 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04803506 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04554528 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24054843 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24054844 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24054781 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04223584 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24054806 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24054814 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04772685 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04566164 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24054963 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24054765 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24054944 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24054903 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24054812 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24054985 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24054825 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24054786 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24054837 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04992888 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24054979 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24054817 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24054919 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24055113 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24054927 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04318054 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24054791 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24054789 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24054778 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24054922 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24054938 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24054965 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04619893 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04636529 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24055118 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04405245 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24054926 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24054961 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24054799 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04537818 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24054853 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24054925 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24054929 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24054876 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24054769 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24054847 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24054982 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24055108 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24055115 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24054852 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24054907 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24054819 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24054776 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24054942 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24054780 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04863920 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04173712 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24054788 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24054902 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04173714 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04685699 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24054958 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04223588 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24054802 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24054980 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24054816 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24054915 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24054883 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24054849 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24054955 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04764259 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04742165 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24055119 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04868803 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24054779 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24054830 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24054905 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24054879 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24054878 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24054839 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24054946 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24054934 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24054954 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24054851 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24054782 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24054991 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24054808 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24054918 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24054981 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04710972 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04803510 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04710381 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04397774 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24054800 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24054857 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24054829 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24054834 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24054797 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24054766 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24054856 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24054937 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24054993 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04613067 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04286821 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24054855 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24054882 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24055112 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24054792 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24054822 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24054832 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24054795 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24054983 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04269106 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24054826 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04383752 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04397773 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24054923 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04173706 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24054892 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04772690 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04318052 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10525528 - -
          Affymetrix Mouse Gene 1.0 ST Array 10525525 - -
          Affymetrix Mouse Gene 1.0 ST Array 10525522 - -
          Affymetrix Mouse Gene 1.0 ST Array 10525534 - -
          Affymetrix Mouse Gene 1.0 ST Array 10525530 - -
          Affymetrix Mouse Gene 1.0 ST Array 10525524 - -
          Affymetrix Mouse Gene 1.0 ST Array 10525526 - -
          Affymetrix Mouse Gene 1.0 ST Array 10525533 - -
          Affymetrix Mouse Gene 1.0 ST Array 10525531 - -
          Affymetrix Mouse Gene 1.0 ST Array 10525527 - -
          Affymetrix Mouse Gene 1.0 ST Array 10525532 - -
          Affymetrix Mouse Gene 1.0 ST Array 10525529 - -
          Affymetrix Mouse Gene 1.0 ST Array 10525535 - -
          Affymetrix Mouse Gene 1.0 ST Array 10525523 - -
          Affymetrix HT MG-430 PM Array Plate 1433062_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1433063_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1430112_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1447215_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10525516 - -
          Mouse Genome 430 2.0 Array 1433063_PM_at - -
          Mouse Genome 430 2.0 Array 1433062_at - -
          Mouse Genome 430 2.0 Array 1430112_at - -
          Mouse Genome 430 2.0 Array 1433062_PM_at - -
          Mouse Genome 430 2.0 Array 1433063_at - -
          Mouse Genome 430 2.0 Array 1430112_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 269701_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 269701_at - -
          Affymetrix Mouse Clariom S Array PSR0500012553.mm.1 TC0500001412.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500012555.mm.1 TC0500001412.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500012562.mm.1 TC0500001412.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500012554.mm.1 TC0500001412.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500012560.mm.1 TC0500001412.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500012563.mm.1 TC0500001412.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500012559.mm.1 TC0500001412.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500012557.mm.1 TC0500001412.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500012561.mm.1 TC0500001412.mm.1 MTA TC
          Mouse Expression Set 430 1433063_PM_at - -
          Mouse Expression Set 430 1430112_PM_at - -
          Mouse Expression Set 430 1433063_at - -
          Mouse Expression Set 430 1433062_at - -
          Mouse Expression Set 430 1430112_at - -
          Mouse Expression Set 430 1433062_PM_at - -
          Murine Genome U74v2 Set 137696_PM_at - -
          Murine Genome U74v2 Set 137696_at - -
          Affymetrix HT Mouse Genome 430B Array 1433062_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1433063_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1433063_at - -
          Affymetrix HT Mouse Genome 430B Array 1430112_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1433062_at - -
          Affymetrix HT Mouse Genome 430B Array 1430112_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1220654 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2423071 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1220654 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline