Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00625905_m1
          See other SDK1 GE Assays ›
          Gene Symbol:
          Sdk1
          Gene Name:
          sidekick homolog 1 (chicken)
          Gene Aliases:
          5330440E10, 6720466O15Rik
          Chromosome Location:
          Chr.5: 141241497 - 142215586 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00625905_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Sdk1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_177879.5 NP_808547.3 9 - 10 1683 70
          GenBank mRNA AK030635.1 - 9 - 10 1194 70
          AK147578.1 - 9 - 10 1728 70
          AY353236.1 - 5 - 6 833 70
          BC060237.1 - 9 - 10 1267 70
          BC138453.1 - 5 - 6 704 8861473 70

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0500001702.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Sdk1

          Gene Ontology Categories:

          Function(s) Process(es)

          cell adhesion
          behavioral response to cocaine
          regulation of dendritic spine development
          molecular_function

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0500015667.mm.1 TC0500001702.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500015667.mm.1 NM_177879.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500015667.mm.1 NM_177879.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0500015672.mm.1 TC0500001702.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500015672.mm.1 NM_177879.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500015672.mm.1 NM_177879.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0500015674.mm.1 TC0500001702.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500015674.mm.1 NM_177879.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500015674.mm.1 NM_177879.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0500015675.mm.1 TC0500001702.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500015675.mm.1 NM_177879.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500015675.mm.1 NM_177879.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0500015676.mm.1 TC0500001702.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500015676.mm.1 NM_177879.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500015676.mm.1 NM_177879.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0500015678.mm.1 TC0500001702.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500015678.mm.1 NM_177879.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500015678.mm.1 NM_177879.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0500015679.mm.1 TC0500001702.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500015679.mm.1 NM_177879.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500015679.mm.1 NM_177879.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0500015662.mm.1 TC0500001702.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500015669.mm.1 TC0500001702.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500015669.mm.1 NM_177879.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500015669.mm.1 NM_177879.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0500015671.mm.1 TC0500001702.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500015671.mm.1 NM_177879.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500015671.mm.1 NM_177879.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0500015673.mm.1 TC0500001702.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500015673.mm.1 NM_177879.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500015673.mm.1 NM_177879.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0500015683.mm.1 TC0500001702.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500015683.mm.1 NM_177879.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500015683.mm.1 NM_177879.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0500015666.mm.1 TC0500001702.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500015666.mm.1 NM_177879.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500015666.mm.1 NM_177879.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0500015681.mm.1 TC0500001702.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500015681.mm.1 NM_177879.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500015681.mm.1 NM_177879.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0500015682.mm.1 TC0500001702.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500015682.mm.1 NM_177879.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500015682.mm.1 NM_177879.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0500015684.mm.1 TC0500001702.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500015684.mm.1 NM_177879.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500015684.mm.1 NM_177879.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0500015670.mm.1 TC0500001702.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500015670.mm.1 NM_177879.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500015670.mm.1 NM_177879.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0500015677.mm.1 TC0500001702.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500015677.mm.1 NM_177879.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500015677.mm.1 NM_177879.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0500015665.mm.1 TC0500001702.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500015665.mm.1 NM_177879.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500015665.mm.1 NM_177879.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0500015680.mm.1 TC0500001702.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500015680.mm.1 NM_177879.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500015680.mm.1 NM_177879.5 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 676744 - -
          ABI Mouse Genome Survey Microarray 706044 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-jwmpg:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0