Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00626135_m1
          See other GALP GE Assays ›
          Gene Symbol:
          Galp
          Gene Name:
          galanin-like peptide
          Gene Aliases:
          -
          Chromosome Location:
          Chr.7: 6197090 - 6216138 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00626135_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Galp
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_178028.2 NP_821171.1 3 - 4 398 59
          GenBank mRNA AF426450.1 - 2 - 3 104 59
          AY163546.1 - 3 - 4 398 59

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700000072.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Galp

          Gene Ontology Categories:

          Function(s) Process(es)

          neuropeptide signaling pathway
          regulation of appetite
          behavioral response to starvation
          defense response to bacterium
          receptor binding
          hormone activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2081652 AY163546.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2081652 NM_178028.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 263908 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 438207 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 885926 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 35220 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 253929 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 606212 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 601690 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 242100 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 207619 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 553357 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 91255 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P125022 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P111382 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2081652 AY163546.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2081652 NM_178028.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P141454 AY163546.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P141454 NM_178028.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24954477 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24954483 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24954486 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24954475 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24954484 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24954498 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24954501 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24954480 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24954478 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24954491 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24954503 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24954494 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24954476 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24954472 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24954499 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24954487 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24954496 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24954492 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24954474 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P111382 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P125022 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06116508 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06116499 - -
          Whole Mouse Genome G4122A A_52_P141454 AY163546.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P141454 NM_178028.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P118865 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P111382 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P125022 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24954523 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24954507 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24954522 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24954552 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24954537 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24954558 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24954531 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24954545 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24954480 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24954548 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24954554 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24954478 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24954496 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24954556 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24954508 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24954525 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24954551 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24954538 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24954571 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24954512 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24954511 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24954534 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24954565 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24954520 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24954481 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24954474 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24954472 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24954484 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24954546 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24954541 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24954550 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24954486 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24954501 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24954535 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24954547 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24954559 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24954517 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24954567 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24954532 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24954473 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24954533 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24954483 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24954563 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24954570 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24954503 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24954487 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24954529 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24954494 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24954553 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24954509 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24954492 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24954498 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06575626 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24954491 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06190912 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24954548 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05912317 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06003183 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05980329 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06634234 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24954550 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24954474 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06222561 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06168290 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24954546 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06594188 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24954484 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06356023 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24954483 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06382751 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06277476 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24954506 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06390132 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24954504 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24954518 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24954486 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24954516 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24954496 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06634235 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24954512 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06371528 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24954498 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05900141 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06115190 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24954525 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24954517 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24954551 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24954473 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24954492 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06222559 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06356026 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06499282 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05909266 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24954477 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24954519 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06305706 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10549824 - -
          Affymetrix Mouse Gene 1.0 ST Array 10549821 - -
          Affymetrix Mouse Gene 1.0 ST Array 10549823 - -
          Affymetrix Mouse Gene 1.0 ST Array 10549822 - -
          Affymetrix Mouse Gene 1.0 ST Array 10549825 - -
          Affymetrix Mouse Gene 1.0 ST Array 10549826 - -
          Affymetrix Mouse Gene 1.1 ST Array 10549820 - -
          Affymetrix Mouse Clariom S Array PSR0700000791.mm.1 TC0700000072.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700000791.mm.1 NM_178028.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700000791.mm.1 NM_178028.2 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 787563 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 3390673 - -
          Illumina mouse-6 v1.0 expression beadchip 3390673 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003390673 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2641914 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2641914 - -
          mouse-6 v1.1 (Illumina) scl0232836.2_13-S - -
          Illumina Sentrix mouse-6.v1 bead array 3390673 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0232836.2_13-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3390673 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2641914 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2641914 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline