Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00626276_m1
          See other 4930486L24RIK GE Assays ›
          Gene Symbol:
          4930486L24Rik
          Gene Name:
          RIKEN cDNA 4930486L24 gene
          Gene Aliases:
          AW545031
          Chromosome Location:
          Chr.13: 60842612 - 60864416 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00626276_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 4930486L24Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_178098.2 NP_835199.1 5 - 6 725 69
          GenBank mRNA AY146988.1 - 5 - 6 725 69
          BC061218.1 - 5 - 6 712 30281034 69

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1300002140.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Protease Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene 4930486L24Rik

          Panther Classification:

          Molecular Function -

          cysteine protease

          Gene Ontology Categories:

          Function(s) Process(es)

          proteolysis
          proteolysis involved in cellular protein catabolic process
          cysteine-type endopeptidase activity
          cysteine-type peptidase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P276479 AY146988.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P276479 BC061218.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P276479 AK145933.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P276479 NM_178098.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P276479 AK015635.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P276479 AY146988.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P276479 BC061218.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P276479 AK145933.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P276479 NM_178098.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P276479 AK015635.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 585386 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 121361 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 29998 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 731858 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 497578 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 747949 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 796712 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 508898 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 325763 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 101995 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 15341 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 33963 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P149478 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P179900 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P276479 AY146988.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P276479 BC061218.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P276479 AK145933.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P276479 NM_178098.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P276479 AK015635.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P276479 AY146988.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P276479 BC061218.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P276479 AK145933.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P276479 NM_178098.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P276479 AK015635.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29087867 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29087848 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29087839 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29087854 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29087870 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29087865 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29087863 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29087856 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29087860 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29087840 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29087850 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29087846 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29087852 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29087836 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29087872 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29087844 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29087833 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29087858 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29087835 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29087842 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29087837 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07896366 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07896383 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P179900 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P149478 - -
          Whole Mouse Genome G4122A A_51_P276479 AY146988.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P276479 BC061218.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P276479 AK145933.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P276479 NM_178098.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P276479 AK015635.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P276479 AY146988.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P276479 BC061218.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P276479 AK145933.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P276479 NM_178098.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P276479 AK015635.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P218440 AY146988.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P218440 BC061218.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P218440 NM_178098.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P105246 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P149478 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P179900 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10724025 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10724030 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10724026 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10724029 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087795 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087895 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087870 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087871 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087813 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087809 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087837 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087753 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087896 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087765 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087836 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087853 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087867 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087792 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087858 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087800 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087823 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087846 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087827 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087820 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087802 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087833 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087778 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087892 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087797 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087854 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087749 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087840 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087746 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087826 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087742 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087759 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087844 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087756 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087798 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087876 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087762 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087828 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087873 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087864 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087806 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087782 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087886 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087767 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087748 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087751 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087874 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087787 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087776 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087824 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087897 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087784 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087815 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087811 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087850 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087741 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087861 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087898 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087818 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087816 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087773 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087764 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087747 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087856 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087878 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087839 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087884 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29087874 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29087844 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29087878 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29087836 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29087867 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29087861 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29087833 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29087839 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10661432 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29087873 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10724032 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29087863 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29087856 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10724025 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29087840 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29087848 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29087864 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29087870 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29087858 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29087850 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29087849 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29087837 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29087854 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10603841 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29087852 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29087876 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10409862 - -
          Affymetrix Mouse Gene 1.0 ST Array 10409865 - -
          Affymetrix Mouse Gene 1.0 ST Array 10409860 - -
          Affymetrix Mouse Gene 1.0 ST Array 10409859 - -
          Affymetrix Mouse Gene 1.0 ST Array 10409861 - -
          Affymetrix Mouse Gene 1.0 ST Array 10409858 - -
          Affymetrix Mouse Gene 1.0 ST Array 10409864 - -
          Affymetrix Mouse Gene 1.0 ST Array 10409863 - -
          Affymetrix HT MG-430 PM Array Plate 1430051_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10409857 - -
          Mouse Genome 430 2.0 Array 1430051_at - -
          Mouse Genome 430 2.0 Array 1430051_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 214639_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 214639_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1300013731.mm.1 NM_178098.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300013731.mm.1 NM_178098.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1300013731.mm.1 TC1300002140.mm.1 MTA TC
          Mouse Expression Set 430 1430051_PM_at - -
          Mouse Expression Set 430 1430051_at - -
          Affymetrix HT Mouse Genome 430B Array 1430051_at - -
          Affymetrix HT Mouse Genome 430B Array 1430051_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 756559 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 6940022 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006940022 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2698334 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2698334 - -
          mouse-6 v1.1 (Illumina) scl0214639.2_15-S - -
          Illumina Sentrix mouse-6.v1 bead array 6940022 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0214639.2_15-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6940022 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2698334 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2698334 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline