Search
Search

Dppa1| Gene Symbol: Dppa1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163358.1 | NP_001156830.1 | 4 - 5 | 276 | 68 | |
| NM_178247.3 | NP_839978.1 | 5 - 6 | 362 | 68 | ||
| GenBank mRNA | AF490345.1 | - | 5 - 6 | 348 | 68 | |
| AK145642.1 | - | 5 - 6 | 363 | 68 | ||
| AK166752.1 | - | 5 - 6 | 360 | 68 | ||
| BC115806.1 | - | 4 - 5 | 303 | 40106955 | 68 | |
| BC115901.1 | - | 4 - 5 | 300 | 40106956 | 68 | |
Set Membership: |
|