Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00649782_gH
          See other BGLAP3 GE Assays ›
          Gene Symbol:
          Bglap3
          Gene Name:
          bone gamma-carboxyglutamate protein 3
          Gene Aliases:
          AI461847, Bglap-rs1, OC-X, ORG, mOC-X
          Chromosome Location:
          Chr.3: 88368615 - 88372749 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00649782_gH
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Bglap3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AA016947.1 - 3 - 4 332 89
          AA475384.1 - 3 - 4 340 89
          AK140913.1 - 5 - 6 1786 89
          BC019774.1 - 6 - 7 767 89
          BC031815.1 - 3 - 4 372 4166503 89
          BC055868.1 - 3 - 4 332 4190595 89
          BC100687.1 - 174 40041102 89
          BC101948.1 - 174 40041107 89
          BC141238.1 - 199 9055850 89
          BC145564.1 - 199 89
          BC145565.1 - 199 9054130 89
          BE625312.1 - 3 - 4 337 89
          BY414395.1 - 126 89

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Inventoried Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2086507 BC145564.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2086507 BC145565.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2086507 BE625312.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2086507 BC141238.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2086507 AK140913.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P101477 BC031815.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P101477 BC055868.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P101477 NM_031368.4 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P101477 AA475384.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2106801 NM_007541.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2106801 AA856059.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2106801 BC019774.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2106801 NM_031368.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2106801 NM_007541.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2106801 AA856059.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2106801 BC019774.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2106801 NM_031368.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2086507 BC145564.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2086507 BC145565.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2086507 BE625312.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2086507 BC141238.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2086507 AK140913.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P101477 BC031815.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P101477 BC055868.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P101477 NM_031368.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P101477 AA475384.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P167410 BC145565.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P167410 BE625312.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P167410 BC141238.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P167410 AK140913.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P167410 BC019774.1 mouse_transcriptome
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22313387 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22313386 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22313365 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22313368 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22313370 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22313366 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22313373 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22313375 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22313391 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22313371 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22313376 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22313378 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22313382 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22313380 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22313418 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10499355 - -
          Affymetrix Mouse Gene 1.0 ST Array 10499361 - -
          Affymetrix Mouse Gene 1.0 ST Array 10499359 - -
          Affymetrix Mouse Gene 1.0 ST Array 10499357 - -
          Affymetrix Mouse Gene 1.0 ST Array 10499360 - -
          Affymetrix Mouse Gene 1.0 ST Array 10499356 - -
          Affymetrix Mouse Gene 1.0 ST Array 10499362 - -
          Affymetrix Mouse Gene 1.1 ST Array 10499358 - -
          Affymetrix Mouse Gene 1.1 ST Array 10499354 - -
          Affymetrix Mouse Clariom S Array PSR0300017471.mm.1 NM_031368.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0300017471.mm.1 TC0300003235.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300017472.mm.1 NM_031368.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0300017472.mm.1 TC0300003235.mm.1 MTA TC
          Mouse Expression Set 430 1449880_s_at - -
          Mouse Expression Set 430 1449880_PM_s_at - -
          Murine Genome U74v2 Set 162296_at - -
          Murine Genome U74v2 Set 100336_PM_s_at - -
          Murine Genome U74v2 Set 100336_s_at - -
          Murine Genome U74v2 Set 162296_PM_at - -
          Murine Genome U74v2 Set 102801_at - -
          Murine Genome U74v2 Set 102801_PM_at - -
          Murine Genome U74v2 Set 169348_PM_at - -
          Murine Genome U74v2 Set 169348_at - -
          Mouse Genome 430A 2.0 Array 1449880_PM_s_at - -
          Mouse Genome 430A 2.0 Array 1449880_s_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 3290397 - -
          Illumina mouse-6 v1.0 expression beadchip 3290397 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003290397 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1233122 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1233122 - -
          mouse-6 v1.1 (Illumina) scl0012097.1_11-S - -
          Illumina Sentrix mouse-6.v1 bead array 3290397 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0012097.1_11-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3290397 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1233122 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1220829 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline