Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00649916_m1
          See other MS4A4B GE Assays ›
          Gene Symbol:
          Ms4a4b
          Gene Name:
          membrane-spanning 4-domains, subfamily A, member 4B
          Gene Aliases:
          AI463180, Chandra, Ly116
          Chromosome Location:
          Chr.19: 11443558 - 11463549 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00649916_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ms4a4b
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_021718.2 NP_068364.1 4 - 5 437 132
          GenBank mRNA AF230640.1 - 4 - 5 132
          AF237913.1 - 4 - 5 406 132
          AK039824.1 - 4 - 5 412 132
          AK088631.1 - 4 - 5 393 132
          AK088780.1 - 4 - 5 443 132
          AK138737.1 - 4 - 5 410 132
          AK172495.1 - 4 - 5 410 132

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1900000219.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ms4a4b

          Panther Classification:

          Molecular Function -

          transporter

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1988975 BC028433.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1988975 AK080934.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1988975 AF354929.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P136294 AK172495.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P136294 AK138737.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P136294 AK088631.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P136294 AK039824.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P136294 NM_021718.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P136294 AK172495.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P136294 AK138737.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P136294 AK088631.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P136294 AK039824.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P136294 NM_021718.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 545058 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 721858 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 713840 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 333575 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 467189 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 801670 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 450613 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 600555 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 762547 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 625333 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 834690 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 938796 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 819588 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 454849 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 91798 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 25070 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 105041 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 167066 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 160990 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 172014 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P170815 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P128618 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P134057 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1988975 BC028433.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1988975 AK080934.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1988975 AF354929.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P136294 AK172495.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P136294 AK138737.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P136294 AK088631.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P136294 AK039824.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P136294 NM_021718.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P136294 AK172495.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P136294 AK138737.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P136294 AK088631.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P136294 AK039824.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P136294 NM_021718.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31956067 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31956060 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31956068 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31956063 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P128618 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09106397 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P134057 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P170815 - -
          Whole Mouse Genome G4122A A_52_P612499 AK037569.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P306949 AK172495.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P306949 BC028433.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P306949 AK138737.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P306949 AK080934.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P306949 AK088631.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P136288 AF230640.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P136294 AK172495.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P136294 AK138737.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P136294 AK088631.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P136294 AK039824.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P136294 NM_021718.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P136294 AK172495.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P136294 AK138737.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P136294 AK088631.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P136294 AK039824.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P136294 NM_021718.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P136289 AF230640.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P128787 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P134057 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P170815 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P128618 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14240655 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31956122 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31956200 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31956232 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31956068 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31956248 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31956174 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31956180 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31956067 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31956128 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31956206 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31956219 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31956106 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31956172 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31956109 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31956107 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31956203 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31956208 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31956239 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31956131 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31956197 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31956063 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31956175 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31956182 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31956245 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31956103 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31956178 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31956092 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31956251 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31956220 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31956230 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31956235 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31956229 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31956170 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31956221 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31956158 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31956079 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14240655 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31956097 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31956068 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31956174 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31956178 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31956063 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31956092 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31956172 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31956077 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31956182 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31956060 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31956175 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31956095 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14327861 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31956082 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31956079 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31956085 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31956180 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31956170 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31956067 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31956086 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31956065 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31956099 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10461611 - -
          Affymetrix Mouse Gene 1.0 ST Array 10461612 - -
          Affymetrix Mouse Gene 1.0 ST Array 10461608 - -
          Affymetrix Mouse Gene 1.0 ST Array 10461606 - -
          Affymetrix Mouse Gene 1.0 ST Array 10461610 - -
          Affymetrix Mouse Gene 1.0 ST Array 10461609 - -
          Affymetrix Mouse Gene 1.0 ST Array 10461607 - -
          Affymetrix Mouse Gene 1.0 ST Array 10461613 - -
          Affymetrix HT MG-430 PM Array Plate 1425325_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1423467_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10461605 - -
          Mouse Genome 430 2.0 Array 1423467_at - -
          Mouse Genome 430 2.0 Array 1423467_PM_at - -
          Mouse Genome 430 2.0 Array 1425325_PM_at - -
          Mouse Genome 430 2.0 Array 1425325_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 60361_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 60361_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1900002720.mm.1 NM_021718.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900002720.mm.1 NM_021718.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1900002720.mm.1 TC1900000219.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900002725.mm.1 NM_021718.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900002725.mm.1 NM_021718.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1900002725.mm.1 TC1900000219.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900002722.mm.1 NM_021718.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900002722.mm.1 NM_021718.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1900002722.mm.1 TC1900000219.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900002737.mm.1 NM_021718.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900002737.mm.1 NM_021718.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1900002737.mm.1 TC1900000219.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900002721.mm.1 NM_021718.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900002721.mm.1 NM_021718.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1900002721.mm.1 TC1900000219.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900002729.mm.1 NM_021718.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900002729.mm.1 NM_021718.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1900002729.mm.1 TC1900000219.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900002723.mm.1 NM_021718.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900002723.mm.1 NM_021718.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1900002723.mm.1 TC1900000219.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900002734.mm.1 NM_021718.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900002734.mm.1 NM_021718.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1900002734.mm.1 TC1900000219.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900002738.mm.1 NM_021718.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900002738.mm.1 NM_021718.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1900002738.mm.1 TC1900000219.mm.1 MTA TC
          Mouse Expression Set 430 1423467_PM_at - -
          Mouse Expression Set 430 1423467_at - -
          Mouse Genome 430A 2.0 Array 1423467_PM_at - -
          Mouse Genome 430A 2.0 Array 1423467_at - -
          Affymetrix HT Mouse Genome 430A Array 1425325_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1423467_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1425325_at - -
          Affymetrix HT Mouse Genome 430A Array 1423467_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 534149 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 103710041 - -
          MouseWG-6 v2 Expression BeadChip 3710154 - -
          Illumina mouse-6 v1.0 expression beadchip 3710154 - -
          Illumina mouse-6 v1.0 expression beadchip 104760152 - -
          Illumina mouse-6 v1.0 expression beadchip 103710041 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003710154 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2599273 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2550935 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2599273 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1241598 - -
          mouse-6 v1.1 (Illumina) scl060361.6_3-S - -
          Illumina Sentrix mouse-6.v1 bead array 104760152 - -
          Illumina Sentrix mouse-6.v1 bead array 3710154 - -
          Illumina Sentrix mouse-6.v1 bead array 103710041 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl060361.6_3-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3710154 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 103710041 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 104760152 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2599273 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1241598 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2550935 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2599273 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-4lm45:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0