Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00650306_g1
          See other NUDT22 GE Assays ›
          Gene Symbol:
          Nudt22
          Gene Name:
          nudix (nucleoside diphosphate linked moiety X)-type motif 22
          Gene Aliases:
          0610006K04Rik, AI851793, AW545547
          Chromosome Location:
          Chr.19: 6993019 - 6996115 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00650306_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Nudt22
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_026675.2 NP_080951.1 2 - 3 529 121
          GenBank mRNA AK002252.1 - 2 - 3 530 121
          BC019768.1 - 1 - 2 506 5124658 121

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1900001029.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Nudt22

          Panther Classification:

          Molecular Function -

          phosphatase

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function
          hydrolase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P436189 NM_026675.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P436189 AK002252.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P436189 BC019768.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P436189 NM_026675.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P436189 AK002252.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P436189 BC019768.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 314634 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 539283 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P127151 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P436189 NM_026675.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P436189 AK002252.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P436189 BC019768.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P436189 NM_026675.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P436189 AK002252.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P436189 BC019768.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31937202 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31937197 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31937220 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31937219 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31937201 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31937199 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31937211 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31937222 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31937218 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31937206 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31937208 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31937209 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31937204 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31937213 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31937221 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P127151 - -
          Whole Mouse Genome G4122A A_51_P436189 NM_026675.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P436189 AK002252.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P436189 BC019768.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P436189 NM_026675.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P436189 AK002252.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P436189 BC019768.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P120251 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P126533 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P127151 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14363983 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14489823 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14489802 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14363982 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14421935 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14421922 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14421931 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31937178 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31937208 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31937171 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31937187 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31937174 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31937152 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31937202 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31937219 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31937209 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31937183 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31937156 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31937155 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31937151 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31937177 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31937218 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31937211 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31937194 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31937201 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31937148 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31937197 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31937143 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31937165 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31937176 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31937146 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31937216 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31937220 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31937180 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31937150 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31937190 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31937158 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31937131 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31937189 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31937199 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31937167 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31937168 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31937222 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31937221 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31937206 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31937213 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31937134 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31937206 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31937219 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31937203 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14421922 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14489817 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31937211 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31937208 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31937201 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31937220 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31937218 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31937204 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31937209 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31937216 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31937197 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31937199 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31937221 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10465583 - -
          Affymetrix Mouse Gene 1.0 ST Array 10465581 - -
          Affymetrix Mouse Gene 1.0 ST Array 10465582 - -
          Affymetrix Mouse Gene 1.0 ST Array 10465585 - -
          Affymetrix Mouse Gene 1.0 ST Array 10465586 - -
          Affymetrix Mouse Gene 1.0 ST Array 10465584 - -
          Affymetrix HT MG-430 PM Array Plate 1451329_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1460358_PM_s_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10465580 - -
          Mouse Genome 430 2.0 Array 1451329_PM_at - -
          Mouse Genome 430 2.0 Array 1460358_s_at - -
          Mouse Genome 430 2.0 Array 1451329_at - -
          Mouse Genome 430 2.0 Array 1460358_PM_s_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 68323_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 68323_at - -
          Affymetrix Mouse Clariom S Array PSR1900009589.mm.1 NM_026675.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900009589.mm.1 NM_026675.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1900009589.mm.1 TC1900001029.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900009591.mm.1 NM_026675.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900009591.mm.1 NM_026675.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1900009591.mm.1 TC1900001029.mm.1 MTA TC
          Mouse Expression Set 430 1451329_PM_at - -
          Mouse Expression Set 430 1460358_s_at - -
          Mouse Expression Set 430 1451329_at - -
          Mouse Expression Set 430 1460358_PM_s_at - -
          Murine Genome U74v2 Set 169758_i_at - -
          Murine Genome U74v2 Set 169758_PM_i_at - -
          Mouse Genome 430A 2.0 Array 1451329_at - -
          Mouse Genome 430A 2.0 Array 1460358_PM_s_at - -
          Mouse Genome 430A 2.0 Array 1460358_s_at - -
          Mouse Genome 430A 2.0 Array 1451329_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1451329_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1460358_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1451329_at - -
          Affymetrix HT Mouse Genome 430A Array 1460358_PM_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 728789 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 2340154 - -
          Illumina mouse-6 v1.0 expression beadchip 2340154 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002340154 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1227723 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1227723 - -
          mouse-6 v1.1 (Illumina) scl068323.1_52-S - -
          Illumina Sentrix mouse-6.v1 bead array 2340154 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl068323.1_52-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2340154 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1227723 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1227723 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl068323.1_52-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline