Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00650523_g1
          See other LCN9 GE Assays ›
          Gene Symbol:
          Lcn9
          Gene Name:
          lipocalin 9
          Gene Aliases:
          9230102I19Rik
          Chromosome Location:
          Chr.2: 25823094 - 25825542 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00650523_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Lcn9
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_029959.2 NP_084235.1 3 - 4 311 142
          GenBank mRNA AF435738.1 - 3 - 4 311 142
          AK020305.1 - 3 - 4 313 142

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200000437.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Lcn9

          Panther Classification:

          Molecular Function -

          transfer/carrier protein

          Gene Ontology Categories:

          Function(s) Process(es)

          transport
          biological_process
          transporter activity
          pheromone binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P618932 AF435738.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P618932 AK020305.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P618932 NM_029959.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P618932 AF435738.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P618932 AK020305.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P618932 NM_029959.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 554086 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 96433 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P113836 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P618932 AF435738.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P618932 AK020305.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P618932 NM_029959.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P618932 AF435738.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P618932 AK020305.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P618932 NM_029959.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P339613 AF435738.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P339613 AK020305.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P339613 NM_029959.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21074415 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21074410 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21074408 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21074422 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21074418 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21074421 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21074428 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21074425 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21074414 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21074411 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P113836 - -
          Whole Mouse Genome G4122A A_52_P618932 AF435738.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P618932 AK020305.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P618932 NM_029959.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P618932 AF435738.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P618932 AK020305.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P618932 NM_029959.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P339613 AF435738.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P339613 AK020305.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P339613 NM_029959.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P102369 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P125280 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P113836 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01854435 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01854442 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21074392 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21074419 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21074418 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21074399 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21074408 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21074359 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21074377 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21074397 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21074370 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21074369 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21074411 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21074364 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21074407 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21074434 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21074362 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21074422 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21074432 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21074428 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21074374 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21074366 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21074421 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21074401 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21074368 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21074415 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21074375 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21074444 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21074382 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21074372 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21074404 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21074390 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21074393 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21074425 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02173321 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21074414 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02173319 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01854437 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21074415 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21074407 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01854435 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21074435 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02078151 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01582866 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01880694 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01880695 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21074390 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21074422 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21074392 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01836734 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02078147 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01854444 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01854454 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01854432 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21074442 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02078156 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01854452 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01984263 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21074428 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01854436 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21074425 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21074411 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21074419 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01854446 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21074447 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02078149 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21074439 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10470209 - -
          Affymetrix Mouse Gene 1.0 ST Array 10470211 - -
          Affymetrix Mouse Gene 1.0 ST Array 10470212 - -
          Affymetrix Mouse Gene 1.0 ST Array 10470210 - -
          Affymetrix Mouse Gene 1.0 ST Array 10470213 - -
          Affymetrix Mouse Gene 1.0 ST Array 10470208 - -
          Affymetrix Mouse Gene 1.0 ST Array 10470207 - -
          Affymetrix HT MG-430 PM Array Plate 1432468_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10470206 - -
          Mouse Genome 430 2.0 Array 1432468_at - -
          Mouse Genome 430 2.0 Array 1432468_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 77704_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 77704_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0200002695.mm.1 NM_029959.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200002695.mm.1 NM_029959.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200002695.mm.1 TC0200000437.mm.1 MTA TC
          Mouse Expression Set 430 1432468_at - -
          Mouse Expression Set 430 1432468_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1432468_at - -
          Affymetrix HT Mouse Genome 430B Array 1432468_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 499129 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 5050450 - -
          Illumina mouse-6 v1.0 expression beadchip 5050450 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005050450 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2734581 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2734581 - -
          mouse-6 v1.1 (Illumina) scl21161.7.1_172-S - -
          Illumina Sentrix mouse-6.v1 bead array 5050450 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl21161.7.1_172-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5050450 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2734581 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2734581 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline