Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00650862_m1
          See other RANBP1 GE Assays ›
          Gene Symbol:
          Ranbp1
          Gene Name:
          RAN binding protein 1
          Gene Aliases:
          Htf9a
          Chromosome Location:
          Chr.16: 18239979 - 18248694 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00650862_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ranbp1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_011239.2 NP_035369.2 3 - 4 447 145
          GenBank mRNA AK002989.1 - 3 - 4 413 145
          AK008276.1 - 3 - 4 410 145
          AK088781.1 - 3 - 4 435 145
          AK151167.1 - 3 - 4 425 145
          AK153074.1 - 3 - 4 429 145
          AK166642.1 - 3 - 4 449 145
          AK167053.1 - 3 - 4 431 145
          BC061140.1 - 3 - 4 466 6775220 145
          L25255.1 - 3 - 4 320 145
          X56045.1 - 3 - 4 452 145
          X56046.1 - 3 - 4 452 145

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1600001308.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ranbp1

          Panther Classification:

          Molecular Function -

          scaffold/adaptor protein

          Gene Ontology Categories:

          Function(s) Process(es)

          spindle organization
          positive regulation of neuron projection development
          positive regulation of mitotic centrosome separation
          intracellular transport
          cellular response to leptomycin B
          GTPase activator activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30578596 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30578595 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30578594 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30578593 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30578598 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2147971 X56046.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2147971 AK153074.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2147971 AK166642.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2147971 AK151167.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2147971 BC061140.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 715671 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 77394 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 116601 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 230048 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 233776 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 61949 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P179544 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2147971 X56046.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2147971 AK153074.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2147971 AK166642.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2147971 AK151167.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2147971 BC061140.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P142465 X56045.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30578634 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30578632 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30578616 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30578627 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30578595 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30578621 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30578601 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30578610 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30578630 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30578603 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30578614 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30578607 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30578622 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30578605 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30578598 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30578609 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30578597 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30578593 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30578618 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30578617 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30578623 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30578629 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30578612 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30578625 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P179544 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08522893 - -
          Whole Mouse Genome G4122A A_51_P142465 X56045.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P106668 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P179544 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12712438 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12712419 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12712422 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12712413 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30578598 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30578595 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30578594 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30578596 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30578593 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30578584 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30578537 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30578569 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30578459 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30578561 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30578499 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30578496 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30578555 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30578545 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30578462 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30578488 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30578467 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30578482 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30578588 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30578547 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30578518 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30578521 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30578590 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30578565 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30578549 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30578502 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30578506 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30578548 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30578585 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30578538 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30578552 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30578550 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30578494 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30578543 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30578525 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30578586 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30578468 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30578539 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30578461 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30578579 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30578557 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30578575 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30578581 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30578465 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30578497 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30578487 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30578474 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30578527 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30578520 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30578573 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30578466 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30578529 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30578564 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30578593 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30578471 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30578509 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30578592 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30578484 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30578541 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30578524 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30578571 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30578470 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30578486 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30578491 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30578562 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30578500 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30578578 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30578584 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30578552 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30578564 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30578593 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12705742 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30578555 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30578561 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12705734 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30578581 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30578569 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30578541 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30578545 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30578547 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30578575 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30578566 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12712422 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30578570 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30578543 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30578586 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30578571 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12655696 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30578554 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30578537 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30578535 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12712432 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30578562 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30578579 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30578536 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30578534 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30578565 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30578559 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30578557 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30578573 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12727797 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30578538 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10438312 - -
          Affymetrix Mouse Gene 1.0 ST Array 10438309 - -
          Affymetrix Mouse Gene 1.0 ST Array 10438310 - -
          Affymetrix Mouse Gene 1.0 ST Array 10438311 - -
          Affymetrix HT MG-430 PM Array Plate 1422547_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10438308 - -
          Mouse Genome 430 2.0 Array 1422547_at - -
          Mouse Genome 430 2.0 Array 1422547_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 19385_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 19385_PM_at - -
          Affymetrix Murine Genome 6500A Array Msa1690_at - -
          Affymetrix Mouse Clariom S Array PSR1600010741.mm.1 TC1600001308.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1600010742.mm.1 NM_011239.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600010742.mm.1 NM_011239.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1600010742.mm.1 TC1600001308.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1600010744.mm.1 NM_011239.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600010744.mm.1 NM_011239.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1600010744.mm.1 TC1600001308.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1600010743.mm.1 NM_011239.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600010743.mm.1 NM_011239.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1600010743.mm.1 TC1600001308.mm.1 MTA TC
          Mouse Expression Set 430 1422547_at - -
          Mouse Expression Set 430 1422547_PM_at - -
          Murine Genome U74v2 Set 98573_r_at - -
          Murine Genome U74v2 Set 98573_PM_r_at - -
          Mouse Genome 430A 2.0 Array 1422547_at - -
          Mouse Genome 430A 2.0 Array 1422547_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1422547_at - -
          Affymetrix HT Mouse Genome 430A Array 1422547_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 579825 - -
          ABI Mouse Genome Survey Microarray 743155 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 430215 - -
          Illumina mouse-6 v1.0 expression beadchip 430215 - -
          Illumina mouse-6 v1.0 expression beadchip 1090180 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000430215 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001090180 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1248082 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2598852 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2598852 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1248082 - -
          mouse-6 v1.1 (Illumina) scl019385.1_25-S - -
          mouse-6 v1.1 (Illumina) scl0019385.1_318-S - -
          Illumina Sentrix mouse-6.v1 bead array 1090180 - -
          Illumina Sentrix mouse-6.v1 bead array 430215 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0019385.1_318-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl019385.1_25-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 430215 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1090180 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2598852 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1248082 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1248082 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2598852 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl019385.1_25-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-5xfkf:80/100.66.78.86:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline