Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00650941_m1
          See other KLRC3 GE Assays ›
          Gene Symbol:
          Klrc3
          Gene Name:
          killer cell lectin-like receptor subfamily C, member 3
          Gene Aliases:
          Klrc2, Nkg2e
          Chromosome Location:
          Chr.6: 129636263 - 129643511 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00650941_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Klrc3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_021378.1 NP_067353.1 2 - 3 271 139
          GenBank mRNA AF195779.1 - 2 - 3 271 139

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0600003242.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Klrc3

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P501382 NM_021378.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P501382 AF195779.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P501382 NM_021378.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P501382 AF195779.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2155146 NM_021378.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2155146 AF195779.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2155146 BC130234.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2155146 BC144931.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2155149 NM_021378.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2155149 AF195779.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 22713 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 125182 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 498524 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P104099 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2155146 NM_021378.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2155146 AF195779.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2155146 BC130234.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2155146 BC144931.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P501382 NM_021378.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P501382 AF195779.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P501382 NM_021378.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P501382 AF195779.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2155149 NM_021378.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2155149 AF195779.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P501382 NM_021378.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P501382 AF195779.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P501382 NM_021378.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P501382 AF195779.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24844702 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24844705 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06068461 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P104099 - -
          Whole Mouse Genome G4122A A_51_P501382 NM_021378.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P501382 AF195779.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P501382 NM_021378.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P501382 AF195779.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P74290 NM_021378.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P74290 AF195779.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P74290 BC130234.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P74290 BC144931.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P122811 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P104099 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24844656 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24844679 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24844673 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24844677 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24844651 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24844676 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24844663 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24844697 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24844665 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24844686 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24844675 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24844655 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24844652 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24844704 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24844685 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24844670 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24844654 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24844705 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24844667 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24844658 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24844682 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24844700 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24844672 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24844683 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24844696 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24844673 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24844682 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24844693 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05659873 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05096651 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24844685 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24844700 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05870887 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24844675 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05763167 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24844705 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24844695 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24844671 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24844697 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24844696 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24844692 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05093818 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24844676 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24844702 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24844672 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05017194 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24844701 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05870890 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05093816 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10548398 - -
          Affymetrix Mouse Gene 1.0 ST Array 10548399 - -
          Affymetrix Mouse Gene 1.0 ST Array 10548400 - -
          Affymetrix HT MG-430 PM Array Plate 1421794_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10548396 - -
          Mouse Genome 430 2.0 Array 1421794_at - -
          Mouse Genome 430 2.0 Array 1421795_s_at - -
          Mouse Genome 430 2.0 Array 1421794_PM_at - -
          Mouse Genome 430 2.0 Array 1421795_PM_s_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 58179_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 58179_at - -
          Affymetrix Mouse Clariom S Array PSR0600025328.mm.1 TC0600003242.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600025328.mm.1 NM_021378.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600025328.mm.1 NM_021378.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600025330.mm.1 TC0600003242.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600025330.mm.1 NM_021378.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600025330.mm.1 NM_021378.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600025331.mm.1 TC0600003242.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600025331.mm.1 NM_021378.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600025331.mm.1 NM_021378.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600025329.mm.1 TC0600003242.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600025329.mm.1 NM_021378.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600025329.mm.1 NM_021378.1 Refseq
          Mouse Expression Set 430 1421795_PM_s_at - -
          Mouse Expression Set 430 1421795_s_at - -
          Mouse Expression Set 430 1421794_at - -
          Mouse Expression Set 430 1421794_PM_at - -
          Mouse Genome 430A 2.0 Array 1421795_PM_s_at - -
          Mouse Genome 430A 2.0 Array 1421794_at - -
          Mouse Genome 430A 2.0 Array 1421795_s_at - -
          Mouse Genome 430A 2.0 Array 1421794_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1421794_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1421794_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 379380 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 3780273 - -
          Illumina mouse-6 v1.0 expression beadchip 3780273 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003780273 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2608368 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2608368 - -
          mouse-6 v1.1 (Illumina) scl28332.3.1_25-S - -
          Illumina Sentrix mouse-6.v1 bead array 3780273 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl28332.3.1_25-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3780273 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2608368 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2608368 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline