| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1800001144.mm.1 |
NM_026301.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800001144.mm.1 |
NM_026301.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800001144.mm.1 |
TC1800000160.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1800001146.mm.1 |
NM_026301.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800001146.mm.1 |
NM_026301.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800001146.mm.1 |
TC1800000160.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1800001145.mm.1 |
NM_026301.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800001145.mm.1 |
NM_026301.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800001145.mm.1 |
TC1800000160.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1800001139.mm.1 |
NM_026301.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800001139.mm.1 |
NM_026301.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800001139.mm.1 |
TC1800000160.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1800001142.mm.1 |
NM_026301.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800001142.mm.1 |
NM_026301.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800001142.mm.1 |
TC1800000160.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1800001143.mm.1 |
NM_026301.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800001143.mm.1 |
NM_026301.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800001143.mm.1 |
TC1800000160.mm.1 |
MTA TC |