Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00651731_m1
          See other CYP2D9 GE Assays ›
          Gene Symbol:
          Cyp2d9
          Gene Name:
          cytochrome P450, family 2, subfamily d, polypeptide 9
          Gene Aliases:
          Cyp2d, P450-2D
          Chromosome Location:
          Chr.15: 82452377 - 82456827 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00651731_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cyp2d9
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_010006.2 NP_034136.2 4 - 5 754 97
          GenBank mRNA AK143963.1 - 4 - 5 754 97
          BC010593.1 - 4 - 5 729 3964178 97
          BC094015.1 - 4 - 5 737 5050233 97
          M23997.1 - 4 - 5 845 97
          M23998.1 - 4 - 5 750 97
          M27168.1 - 4 - 5 729 97

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1500000757.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection P450 Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cyp2d9

          Panther Classification:

          Molecular Function -

          oxidoreductase oxygenase metabolite interconversion enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          xenobiotic metabolic process
          arachidonic acid metabolic process
          exogenous drug catabolic process
          oxidation-reduction process
          monooxygenase activity
          iron ion binding
          arachidonic acid epoxygenase activity
          steroid hydroxylase activity
          oxidoreductase activity
          oxidoreductase activity, acting on paired donors, with incorporation or reduction of molecular oxygen
          oxidoreductase activity, acting on paired donors, with incorporation or reduction of molecular oxygen, reduced flavin or flavoprotein as one donor, and incorporation of one atom of oxygen
          oxygen binding
          heme binding
          metal ion binding
          aromatase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P128963 M23998.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P128963 M27168.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P128963 BC010593.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P128963 BC094015.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P128963 M23997.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 29042 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 556806 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P107247 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P128963 M23998.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P128963 M27168.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P128963 BC010593.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P128963 BC094015.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P128963 M23997.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P478303 M23998.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P478303 M27168.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P478303 BC010593.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P478303 BC061505.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P478303 BC094015.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P124133 NR_033515.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P124133 BC059104.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P124133 BC016256.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P124133 BC025224.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P124133 AK090296.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30383433 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30383481 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30383441 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30383443 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30383440 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30383466 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30383479 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30383444 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30383435 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P107247 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03084072 - -
          Whole Mouse Genome G4122A A_51_P124133 NR_033515.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P124133 BC059104.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P124133 BC016256.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P124133 BC025224.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P124133 AK090296.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P478303 M23998.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P478303 M27168.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P478303 BC010593.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P478303 BC061505.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P478303 BC094015.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P128963 M23998.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P128963 M27168.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P128963 BC010593.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P128963 BC094015.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P128963 M23997.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P107247 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12446961 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30383499 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30383500 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30383411 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30383487 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30383440 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30383535 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30383479 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30383529 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30383533 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30383526 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30383443 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30383498 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30383441 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30383508 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30383542 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30383538 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30383530 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30383484 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30383540 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30383515 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30383438 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30383497 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30383544 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30383552 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30383486 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12360473 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30383444 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30383497 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30383438 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11900801 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30383418 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30383533 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30383525 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30383443 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30383542 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30383546 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30383481 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11890219 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30383461 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12226253 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30383499 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30383538 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30383508 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30383487 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30383490 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30383536 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11890214 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30383408 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30383529 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30383421 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12360478 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12123132 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30383498 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30383405 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30383466 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12360485 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30383535 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12004320 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30383479 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12062993 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30383402 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30383435 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30383441 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30383530 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30383411 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30383440 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12360492 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30383414 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30383484 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10425770 - -
          Affymetrix Mouse Gene 1.0 ST Array 10425765 - -
          Affymetrix Mouse Gene 1.0 ST Array 10425771 - -
          Affymetrix Mouse Gene 1.0 ST Array 10425767 - -
          Affymetrix Mouse Gene 1.0 ST Array 10425769 - -
          Affymetrix Mouse Gene 1.0 ST Array 10425768 - -
          Affymetrix Mouse Gene 1.0 ST Array 10425764 - -
          Affymetrix Mouse Gene 1.0 ST Array 10425766 - -
          Affymetrix HT MG-430 PM Array Plate 1419349_PM_a_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10425763 - -
          Mouse Genome 430 2.0 Array 1419349_PM_a_at - -
          Mouse Genome 430 2.0 Array 1419349_a_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 13105_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 13105_PM_at - -
          Affymetrix Murine Genome 6500A Array Msa10470_at - -
          Affymetrix Mouse Clariom S Array PSR1500005955.mm.1 NM_010006.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500005955.mm.1 NM_010006.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1500005955.mm.1 TC1500000757.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500005958.mm.1 NM_010006.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500005958.mm.1 NM_010006.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1500005958.mm.1 TC1500000757.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500005961.mm.1 NM_010006.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500005961.mm.1 NM_010006.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1500005961.mm.1 TC1500000757.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500005953.mm.1 NM_010006.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500005953.mm.1 NM_010006.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1500005953.mm.1 TC1500000757.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500005954.mm.1 NM_010006.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500005954.mm.1 NM_010006.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1500005954.mm.1 TC1500000757.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500005956.mm.1 NM_010006.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500005956.mm.1 NM_010006.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1500005956.mm.1 TC1500000757.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500005960.mm.1 NM_010006.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500005960.mm.1 NM_010006.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1500005960.mm.1 TC1500000757.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500005952.mm.1 NM_010006.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500005952.mm.1 NM_010006.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1500005952.mm.1 TC1500000757.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500005957.mm.1 NM_010006.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500005957.mm.1 NM_010006.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1500005957.mm.1 TC1500000757.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500005959.mm.1 NM_010006.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500005959.mm.1 NM_010006.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1500005959.mm.1 TC1500000757.mm.1 MTA TC
          Mouse Expression Set 430 1419349_PM_a_at - -
          Mouse Expression Set 430 1419349_a_at - -
          Murine Genome U74v2 Set 94539_PM_f_at - -
          Murine Genome U74v2 Set 162174_PM_at - -
          Murine Genome U74v2 Set 94539_f_at - -
          Murine Genome U74v2 Set 162174_at - -
          Mouse Genome 430A 2.0 Array 1419349_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1419349_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1419349_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1419349_PM_a_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 575431 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 2940133 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002940133 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2687507 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2687507 - -
          mouse-6 v1.1 (Illumina) scl47686.9.1_164-S - -
          Illumina Sentrix mouse-6.v1 bead array 2940133 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl47686.9.1_164-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2940133 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2687507 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2687507 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline