Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00652739_s1
          See other VMN1R74 GE Assays ›
          Gene Symbol:
          Vmn1r74
          Gene Name:
          vomeronasal 1 receptor 74
          Gene Aliases:
          V1rg5
          Chromosome Location:
          Chr.7: 11846775 - 11847689 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00652739_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Vmn1r74
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_134206.1 NP_598967.1 1 341 153

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Vmn1r74

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          response to pheromone
          pheromone binding
          pheromone receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P332506 NM_134206.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P332506 NM_134206.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P122051 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P332506 NM_134206.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P332506 NM_134206.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P332506 NM_134206.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P332506 NM_134206.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24969805 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24969798 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24969787 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24969788 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24969784 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24969780 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24969790 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24969799 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24969802 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24969785 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24969789 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24969794 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24969792 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24969782 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P122051 - -
          Whole Mouse Genome G4122A A_51_P332506 NM_134206.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P332506 NM_134206.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P114965 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P137093 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P122051 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06617861 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06002024 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06002026 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24969820 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24969766 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24969811 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24969843 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24969815 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24969835 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24969816 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24969818 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24969805 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24969838 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24969813 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24969768 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24969801 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24969824 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24969842 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24969848 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24969780 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24969785 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24969784 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24969830 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24969782 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24969799 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24969792 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24969845 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24969829 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24969787 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24969764 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24969812 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24969809 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24969806 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24969790 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24969831 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24969776 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24969794 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24969840 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24969828 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24969826 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24969805 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24969788 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24969780 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24969812 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24969808 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24969838 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24969785 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24969789 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24969787 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24969840 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24969807 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06247872 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24969782 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24969820 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24969829 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24969826 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06501522 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24969818 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24969830 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24969813 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06058709 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24969835 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24969809 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24969794 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24969831 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24969824 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24969776 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24969833 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06643319 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24969816 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24969798 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24969790 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24969806 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24969784 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24969837 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24969799 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06165286 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24969815 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06002022 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24969792 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24969811 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24969828 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10549985 - -
          Affymetrix Mouse Gene 1.1 ST Array 10549984 - -
          Affymetrix Mouse Clariom S Array PSR0700001031.mm.1 TC0700000120.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700001031.mm.1 NM_134206.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700001031.mm.1 NM_134206.1 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 538961 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 2060692 - -
          Illumina mouse-6 v1.0 expression beadchip 101570438 - -
          Illumina mouse-6 v1.0 expression beadchip 2060692 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002060692 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2463192 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2463192 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1230014 - -
          mouse-6 v1.1 (Illumina) GI_21717734-S - -
          mouse-6 v1.1 (Illumina) scl9723.1.1_216-S - -
          Illumina Sentrix mouse-6.v1 bead array 2060692 - -
          Illumina Sentrix mouse-6.v1 bead array 101570438 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl9723.1.1_216-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 101570438 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2060692 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2838831 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2838831 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline