Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00653204_s1
          See other OLFR114 GE Assays ›
          Gene Symbol:
          Olfr114
          Gene Name:
          olfactory receptor 114
          Gene Aliases:
          MOR218-10
          Chromosome Location:
          Chr.17: 37589413 - 37590351 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00653204_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr114
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146287.1 NP_666399.1 1 549 150
          GenBank mRNA BC132209.1 - 1 579 40130486 150
          BC132573.1 - 1 579 40130850 150

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr114

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          G-protein coupled receptor activity
          olfactory receptor activity
          odorant binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1981135 BC132209.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1981135 BC132573.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1981135 NM_146287.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P175415 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1981135 BC132209.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1981135 BC132573.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1981135 NM_146287.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P434928 BC132209.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P434928 BC132573.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P434928 NM_146287.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31169284 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31169267 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31169266 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31169289 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31169304 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31169306 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31169265 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31169292 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31169305 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31169294 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31169288 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31169277 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31169296 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31169268 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31169300 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31169291 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31169287 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P175415 - -
          Whole Mouse Genome G4122A A_51_P434928 BC132209.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P434928 BC132573.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P434928 NM_146287.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P122264 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P175415 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13482691 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13482694 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31169266 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31169305 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31169284 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31169291 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31169296 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31169268 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31169267 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31169302 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31169294 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31169304 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31169288 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31169287 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31169284 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31169294 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13482701 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13482695 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31169305 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31169266 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31169277 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13101376 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31169268 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31169289 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31169265 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31169302 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31169296 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13518211 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31169287 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13482710 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31169271 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31169306 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31169291 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31169288 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31169292 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10450896 - -
          Affymetrix Mouse Gene 1.1 ST Array 10450895 - -
          Affymetrix Mouse Clariom S Array PSR1700019607.mm.1 TC1700002075.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700019605.mm.1 TC1700002075.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700019606.mm.1 NM_146287.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700019606.mm.1 NM_146287.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700019606.mm.1 TC1700002075.mm.1 MTA TC
          Murine Genome U74v2 Set 133989_PM_at - -
          Murine Genome U74v2 Set 133989_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 878080 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 730095 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000730095 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2616202 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2616202 - -
          mouse-6 v1.1 (Illumina) scl7847.1.1_328-S - -
          Illumina Sentrix mouse-6.v1 bead array 730095 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl7847.1.1_328-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 730095 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2616202 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2616202 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline