Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00653255_s1
          See other OLFR446 GE Assays ›
          Gene Symbol:
          Olfr446
          Gene Name:
          olfactory receptor 446
          Gene Aliases:
          MOR261-12
          Chromosome Location:
          Chr.6: 42927233 - 42928159 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00653255_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr446
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146295.1 NP_666407.1 1 751 142

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr446

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2069674 NM_146295.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 587134 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 217349 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 180595 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 268184 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 61563 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 288779 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 789471 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 294410 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 168516 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 140330 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2069674 NM_146295.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P514902 NM_146295.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24400753 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01314792 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05879365 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05879344 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01314799 - -
          Whole Mouse Genome G4122A A_51_P514902 NM_146295.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P131157 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P05879344 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05793570 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05407211 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05407218 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05407217 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05407215 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05793573 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400738 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400741 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24401092 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24401091 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400753 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24400740 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24401437 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P33008363 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400738 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05097353 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P33008366 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24401435 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24401515 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05420070 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24401519 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400743 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05097354 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24401499 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05069081 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P33008361 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400753 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24401433 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24401432 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24401448 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24401511 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24401089 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05420092 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400751 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24401500 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05069075 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05381862 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05420090 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05268649 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05420069 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24401088 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24401424 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24401092 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24401517 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24400739 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24401450 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24401498 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24401091 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10537816 - -
          Affymetrix Mouse Gene 1.0 ST Array 10537808 - -
          Affymetrix Mouse Gene 1.0 ST Array 10537815 - -
          Affymetrix Mouse Gene 1.0 ST Array 10537813 - -
          Affymetrix Mouse Gene 1.0 ST Array 10537812 - -
          Affymetrix Mouse Gene 1.0 ST Array 10537814 - -
          Affymetrix Mouse Gene 1.1 ST Array 10537807 - -
          Affymetrix Mouse Gene 1.1 ST Array 10537811 - -
          Affymetrix Mouse Clariom S Array PSR0600003743.mm.1 TC0600000473.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600003743.mm.1 NM_146295.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600003743.mm.1 NM_146295.1 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 894548 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 2060333 - -
          Illumina mouse-6 v1.0 expression beadchip 2060333 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002060333 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2716580 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2716580 - -
          mouse-6 v1.1 (Illumina) scl0258292.1_115-S - -
          Illumina Sentrix mouse-6.v1 bead array 2060333 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0258292.1_115-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2060333 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2716580 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2716580 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline