Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00653289_s1
          See other OLFR510 GE Assays ›
          Gene Symbol:
          Olfr510
          Gene Name:
          olfactory receptor 510
          Gene Aliases:
          MOR204-34
          Chromosome Location:
          Chr.7: 108666424 - 108668362 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00653289_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr510
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146311.1 NP_666423.1 1 242 98

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr510

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          signal transduction
          G-protein coupled receptor signaling pathway
          sensory perception of smell
          response to stimulus
          detection of chemical stimulus involved in sensory perception of smell
          signal transducer activity
          G-protein coupled receptor activity
          olfactory receptor activity
          odorant binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P148478 NM_146311.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P148478 NM_146311.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P131799 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P148478 NM_146311.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P148478 NM_146311.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P148478 NM_146311.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P148478 NM_146311.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25404172 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25404183 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25404181 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25404202 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25404193 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25404186 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25404199 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25404187 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25404176 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25404178 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25404197 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25404179 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25404185 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25404175 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25404173 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25404188 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P131799 - -
          Whole Mouse Genome G4122A A_51_P148478 NM_146311.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P148478 NM_146311.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P133611 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P131799 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06307935 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06307919 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06307941 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06307916 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06307914 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25404169 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25404166 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25404197 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25404202 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25404175 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25404205 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25404168 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25404187 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25404173 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25404192 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25404190 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25404179 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25404183 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25404199 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25404181 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25404170 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25404186 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25404202 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25404178 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06441838 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25404207 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25404173 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25404175 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25404186 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25404206 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25404197 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25404190 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25404181 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25404183 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25404193 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05888339 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06441841 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25404187 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25404199 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25404188 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25404192 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25404179 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25404205 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10556161 - -
          Affymetrix Mouse Gene 1.1 ST Array 10556160 - -
          Affymetrix Mouse Clariom S Array PSR0700013288.mm.1 NM_146311.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700013288.mm.1 NM_146311.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700013288.mm.1 TC0700001603.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 650588 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 730091 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000730091 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1225331 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1225331 - -
          mouse-6 v1.1 (Illumina) scl32212.1.1_45-S - -
          Illumina Sentrix mouse-6.v1 bead array 730091 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl32212.1.1_45-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 730091 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1225331 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1225331 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-hznln:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline