Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00653401_s1
          See other OLFR390 GE Assays ›
          Gene Symbol:
          Olfr390
          Gene Name:
          olfactory receptor 390
          Gene Aliases:
          MOR135-26
          Chromosome Location:
          Chr.11: 73786940 - 73787875 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00653401_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr390
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146347.1 NP_666459.1 1 56 74

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr390

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception
          detection of chemical stimulus involved in sensory perception of smell
          transmembrane signaling receptor activity
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2024720 NM_146347.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2024720 NM_146347.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P487379 NM_146347.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27918053 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27918050 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27918049 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27918048 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27918042 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27918043 - -
          Whole Mouse Genome G4122A A_52_P487379 NM_146347.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P117112 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P103782 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27917990 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27918030 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27918061 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27917971 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27918005 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27917976 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27918059 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27918042 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27917985 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27917981 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27917980 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27917975 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27917988 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27918010 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27918053 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27918049 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27917996 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27918056 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27918018 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27918065 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27917989 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27917965 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27918038 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27917978 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27918017 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27917973 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27917987 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27917993 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27917967 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27918027 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27918033 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27917979 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27918070 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27918057 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27918025 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27918021 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27918055 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27917983 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27917999 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27918024 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27917995 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27918015 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27917992 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27917997 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27918068 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27918003 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27918063 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27918069 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27917968 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27918050 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27918013 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27918058 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27918055 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27918030 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27918047 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09366039 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27918025 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27918027 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27918005 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09304870 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09610792 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27918057 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09171165 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27918038 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27918042 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27918043 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27918049 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27918050 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27918028 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27918033 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09171169 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27918024 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27918061 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27918010 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27918015 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27918017 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27918021 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27918063 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09625572 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27918018 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27918059 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27918053 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27918020 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10378408 - -
          Affymetrix Mouse Gene 1.1 ST Array 10378407 - -
          Affymetrix Mouse Clariom S Array PSR1100009348.mm.1 NM_146347.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100009348.mm.1 NM_146347.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100009348.mm.1 TC1100001028.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 428759 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 1410692 - -
          Illumina mouse-6 v1.0 expression beadchip 1410692 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001410692 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2766903 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2766903 - -
          mouse-6 v1.1 (Illumina) scl14051.1.1_330-S - -
          Illumina Sentrix mouse-6.v1 bead array 1410692 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl14051.1.1_330-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1410692 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2820869 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2820869 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline