Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00653424_s1
          See other OLFR706 GE Assays ›
          Gene Symbol:
          Olfr706
          Gene Name:
          olfactory receptor 706
          Gene Aliases:
          MOR283-11
          Chromosome Location:
          Chr.7: 106885865 - 106886815 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00653424_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr706
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146353.2 NP_666465.1 1 344 161

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor Detect Genomic DNA Recommended Assay Mus musculus Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr706

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P232649 NM_146353.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P232649 NM_146353.2 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P161496 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P232649 NM_146353.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P232649 NM_146353.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P232649 NM_146353.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P232649 NM_146353.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25397009 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25397017 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25396998 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25397025 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25397022 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25396996 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25397030 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25397006 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25397032 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25397010 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25397005 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25397019 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25396997 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25397033 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25397035 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25397015 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25397013 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25397007 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25397024 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25397031 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25397003 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25397018 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25397002 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25397004 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25397000 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25397008 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25397014 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25397037 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25397026 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P161496 - -
          Whole Mouse Genome G4122A A_51_P232649 NM_146353.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P232649 NM_146353.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P130364 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P161496 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05997266 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05997258 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05908374 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05908360 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05997253 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05997255 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396996 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25397070 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25397024 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396970 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25397033 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25397059 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25397025 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25397049 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396951 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25397015 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396998 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396956 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25397008 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25397055 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25397065 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25397054 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396967 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25397002 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396964 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25397074 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25397072 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25397063 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25397057 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25397051 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25397003 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25397001 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25397005 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396958 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25397017 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25397068 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25397047 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396953 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25397022 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396957 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25397026 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25397061 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25397030 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25397035 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396962 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396959 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396995 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25397032 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25397037 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396972 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25397006 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25397013 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25397019 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396950 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25397007 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25397026 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25397035 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05997258 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25397031 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05946653 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25397005 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25396998 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06486739 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25397003 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25397001 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05946645 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25397002 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25397004 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25397022 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25397010 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05997256 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25397018 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25397030 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25397000 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25397019 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25397015 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25397006 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05908378 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25397017 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25396995 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25396996 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05908373 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25397013 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25397012 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25397007 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10566610 - -
          Affymetrix Mouse Gene 1.1 ST Array 10566609 - -
          Affymetrix Mouse Clariom S Array PSR0700033456.mm.1 NM_146353.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700033456.mm.1 NM_146353.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700033456.mm.1 TC0700004023.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 1990092 - -
          Illumina mouse-6 v1.0 expression beadchip 1990092 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001990092 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2621223 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2621223 - -
          mouse-6 v1.1 (Illumina) scl8844.1.1_9-S - -
          Illumina Sentrix mouse-6.v1 bead array 1990092 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl8844.1.1_9-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1990092 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2621223 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2621223 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline