Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00653594_s1
          See other OLFR837 GE Assays ›
          Gene Symbol:
          Olfr837
          Gene Name:
          olfactory receptor 837
          Gene Aliases:
          MOR152-2
          Chromosome Location:
          Chr.9: 19136928 - 19137967 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00653594_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr837
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146565.2 NP_666776.1 1 413 145
          GenBank mRNA BC140981.1 - 1 413 9055593 145

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr837

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P273064 NM_146565.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P273064 BC140981.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P273064 NM_146565.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P273064 BC140981.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 175321 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P119621 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P273064 NM_146565.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P273064 BC140981.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P273064 NM_146565.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P273064 BC140981.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P273064 NM_146565.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P273064 BC140981.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P273064 NM_146565.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P273064 BC140981.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26339296 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26339274 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26339273 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26339280 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26339275 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26339270 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26339297 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26339300 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26339285 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26339303 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26339278 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26339277 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26339282 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26339301 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26339287 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P119621 - -
          Whole Mouse Genome G4122A A_51_P273064 NM_146565.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P273064 BC140981.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P273064 NM_146565.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P273064 BC140981.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P119621 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08185517 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08185540 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08185531 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08185525 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08185532 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26339305 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26339271 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26339282 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26339309 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26339300 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26339277 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26339297 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26339273 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26339307 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26339285 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26339275 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26339278 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26339274 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26339286 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26339303 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26339268 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26339280 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26339282 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26339311 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26339280 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26339268 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26339277 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26339305 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26339275 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26339285 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26339300 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26339296 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26339274 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26339278 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26339298 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26339269 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26339273 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07541022 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26339307 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26339303 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26339286 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26339306 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26339302 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08185539 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26339309 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26339297 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10583391 - -
          Affymetrix Mouse Gene 1.1 ST Array 10583390 - -
          Affymetrix Mouse Clariom S Array PSR0900000971.mm.1 NM_146565.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900000971.mm.1 NM_146565.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0900000971.mm.1 TC0900000146.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900000970.mm.1 NM_146565.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900000970.mm.1 NM_146565.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0900000970.mm.1 TC0900000146.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 415481 - -
          ABI Mouse Genome Survey Microarray 415566 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 670528 - -
          Illumina mouse-6 v1.0 expression beadchip 670528 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000670528 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2643659 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2643659 - -
          mouse-6 v1.1 (Illumina) scl37255.1.1_329-S - -
          Illumina Sentrix mouse-6.v1 bead array 670528 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl37255.1.1_329-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 670528 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2643659 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2643659 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline