Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00653604_s1
          See other OLFR830 GE Assays ›
          Gene Symbol:
          Olfr830
          Gene Name:
          olfactory receptor 830
          Gene Aliases:
          MOR152-1
          Chromosome Location:
          Chr.9: 18874960 - 18876276 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00653604_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr830
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146566.1 NP_666777.1 1 353 148

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr830

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2014144 NM_146566.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P134712 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2014144 NM_146566.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P185161 NM_146566.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26338621 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26338615 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26338646 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26338625 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26338629 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26338628 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26338642 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26338617 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26338633 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26338639 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26338643 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26338613 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26338618 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26338619 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26338612 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26338622 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26338645 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26338641 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26338623 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26338611 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26338631 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P134712 - -
          Whole Mouse Genome G4122A A_51_P185161 NM_146566.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P108540 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P134712 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07879322 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07879339 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07879341 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07879345 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07879353 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26338622 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26338617 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26338643 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26338651 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26338621 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26338629 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26338659 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26338623 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26338611 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26338647 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26338619 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26338636 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26338616 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26338645 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26338656 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26338655 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26338657 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26338663 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26338631 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26338625 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26338650 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26338666 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26338613 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26338618 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26338642 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26338648 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26338639 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26338617 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07879344 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26338648 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26338651 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26338650 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26338656 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26338639 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07501966 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26338623 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26338613 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26338647 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26338611 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26338629 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26338621 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26338641 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26338622 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26338657 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26338655 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26338653 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26338642 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26338618 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26338643 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26338616 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26338636 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26338612 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26338659 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26338619 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26338628 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26338631 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08075795 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26338633 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26338661 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26338615 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10583379 - -
          Affymetrix Mouse Gene 1.1 ST Array 10583378 - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 687394 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 5860452 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005860452 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2715382 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2715382 - -
          mouse-6 v1.1 (Illumina) scl7640.1.1_312-S - -
          Illumina Sentrix mouse-6.v1 bead array 5860452 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl7640.1.1_312-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5860452 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2715382 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2715382 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-4lm45:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0