Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00653627_s1
          See other OLFR703 GE Assays ›
          Gene Symbol:
          Olfr703
          Gene Name:
          olfactory receptor 703
          Gene Aliases:
          MOR283-7
          Chromosome Location:
          Chr.7: 106843583 - 106845572 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00653627_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr703
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146596.1 NP_666807.1 1 657 132
          GenBank mRNA BC111815.1 - 1 656 132

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr703

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P281273 BC111815.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P281273 NM_146596.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P281273 BC111815.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P281273 NM_146596.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 356045 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 447019 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 233029 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 913039 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 894251 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 56290 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 143018 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 21577 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 58978 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 23521 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P163968 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P281273 BC111815.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P281273 NM_146596.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P281273 BC111815.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P281273 NM_146596.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P281273 BC111815.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P281273 NM_146596.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P281273 BC111815.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P281273 NM_146596.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P163968 - -
          Whole Mouse Genome G4122A A_51_P281273 BC111815.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P281273 NM_146596.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P281273 BC111815.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P281273 NM_146596.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P103941 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P163968 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396841 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396849 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396847 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396831 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396838 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396835 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396843 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396839 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396853 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396840 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396860 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396846 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396861 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396851 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396864 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25396833 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06115211 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25396838 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25396853 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25396849 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06115209 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25396837 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25396846 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25396864 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06115196 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25396851 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25396832 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25396847 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25396833 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25396835 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25396861 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25396831 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25396860 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25396841 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25396839 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25396863 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25396840 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10556030 - -
          Affymetrix Mouse Gene 1.1 ST Array 10556029 - -
          Affymetrix Mouse Clariom S Array PSR0700013156.mm.1 TC0700001565.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700013156.mm.1 NM_146596.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700013156.mm.1 NM_146596.1 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 642169 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 1500546 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001500546 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1244304 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1244304 - -
          mouse-6 v1.1 (Illumina) scl32231.1.1_325-S - -
          Illumina Sentrix mouse-6.v1 bead array 1500546 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl32231.1.1_325-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1500546 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1244304 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1244304 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline