Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00653650_s1
          See other OLFR1445 GE Assays ›
          Gene Symbol:
          Olfr1445
          Gene Name:
          olfactory receptor 1445
          Gene Aliases:
          MOR202-7
          Chromosome Location:
          Chr.19: 12883883 - 12884827 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00653650_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr1445
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146699.1 NP_666910.1 1 904 73
          GenBank mRNA BC138186.1 - 1 932 8861205 73

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr1445

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          G-protein coupled receptor activity
          olfactory receptor activity
          odorant binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1964692 BC138186.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1964692 NM_146699.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P111286 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1964692 BC138186.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1964692 NM_146699.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P417568 BC138186.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P417568 NM_146699.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31962912 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31962879 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31962878 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31962888 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31962881 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31962914 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31962895 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31962908 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31962871 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31962880 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31962893 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31962916 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31962873 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31962885 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31962892 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31962887 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31962917 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31962865 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31962907 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31962868 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31962909 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31962866 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31962913 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31962883 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31962905 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31962876 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31962869 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P111286 - -
          Whole Mouse Genome G4122A A_51_P417568 BC138186.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P417568 NM_146699.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P127193 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P111286 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14286129 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14286132 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14286133 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14287291 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31962957 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31962938 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31962869 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31962878 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31962965 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31962885 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31962868 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31962939 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31962916 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31962880 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31962947 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31962881 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31962923 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31962864 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31962907 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31962912 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31962905 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31962909 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31962866 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31962871 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31962887 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31962876 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31962944 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31962895 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31962873 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31962908 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31962967 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31962962 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31962883 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31962914 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31962879 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31962892 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31962959 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31962942 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31962912 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31962871 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31962892 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31962907 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31962868 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31962897 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31962917 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31962876 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31962862 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31962914 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31962866 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14466047 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31962908 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31962879 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31962873 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31962903 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14286141 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31962916 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31962881 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31962909 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31962887 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31962885 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31962869 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31962895 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14274280 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31962889 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31962891 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31962880 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31962883 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14286129 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31962864 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31962878 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10461781 - -
          Affymetrix Mouse Gene 1.1 ST Array 10461780 - -
          Affymetrix Mouse Clariom S Array PSR1900003019.mm.1 TC1900000258.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 897585 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 2450064 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002450064 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2685864 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2685864 - -
          mouse-6 v1.1 (Illumina) scl6294.1.1_46-S - -
          Illumina Sentrix mouse-6.v1 bead array 2450064 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl6294.1.1_46-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2450064 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2685864 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2685864 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline