Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00653875_s1
          See other OLFR1406 GE Assays ›
          Gene Symbol:
          Olfr1406
          Gene Name:
          olfactory receptor 1406
          Gene Aliases:
          MOR267-5
          Chromosome Location:
          Chr.1: 173183442 - 173184504 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00653875_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr1406
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146763.2 NP_666974.2 1 706 115
          GenBank mRNA BC139449.1 - 1 706 8862471 115
          BC139452.1 - 1 706 8862474 115

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr1406

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception
          detection of chemical stimulus involved in sensory perception of smell
          transmembrane signaling receptor activity
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2151950 BC139449.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2151950 BC139452.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P124230 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2151950 BC139449.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2151950 BC139452.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P212135 NM_146763.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20836574 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20836585 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20836591 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20836581 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20836608 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20836594 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20836610 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20836607 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20836595 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20836597 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20836605 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20836577 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20836579 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20836602 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20836613 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20836614 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20836599 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20836592 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20836576 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P124230 - -
          Whole Mouse Genome G4122A A_51_P212135 NM_146763.2 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P124230 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00524005 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00672955 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00672959 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00672960 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20836616 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20836553 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20836608 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20836634 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20836640 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20836605 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20836610 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20836642 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20836566 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20836638 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20836599 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20836646 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20836576 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20836644 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20836579 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20836625 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20836607 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20836619 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20836602 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20836645 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20836562 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20836647 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20836597 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20836613 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20836577 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20836631 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20836595 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20836563 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20836574 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20836621 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20836636 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20836567 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20836561 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20836648 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20836557 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20836623 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20836572 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20836569 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20836565 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20836627 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20836608 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20836565 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00523998 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20836613 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00523993 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01089449 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20836631 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00952625 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00521097 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20836593 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20836625 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20836594 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20836607 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00521095 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20836600 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00524001 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20836561 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20836581 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20836599 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20836592 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00521086 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20836567 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00952622 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20836628 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20836621 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20836602 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20836627 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00523999 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00523988 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20836595 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01089450 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20836614 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00523996 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20836569 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20836585 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01089456 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20836597 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20836583 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20836574 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20836616 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00952623 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20836618 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20836572 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20836562 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10360337 - -
          Affymetrix Mouse Gene 1.1 ST Array 10360336 - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 402448 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 2230435 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002230435 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2769151 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2769151 - -
          mouse-6 v1.1 (Illumina) scl0258758.1_45-S - -
          Illumina Sentrix mouse-6.v1 bead array 2230435 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0258758.1_45-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2230435 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2957081 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2957081 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2769151 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-jwmpg:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0