Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00654451_s1
          See other OLFR1126 GE Assays ›
          Gene Symbol:
          Olfr1126
          Gene Name:
          olfactory receptor 1126
          Gene Aliases:
          MOR264-5
          Chromosome Location:
          Chr.2: 87453962 - 87458111 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00654451_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr1126
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146837.2 NP_667048.2 1 638 120

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr1126

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P143040 NM_146837.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P143040 NM_146837.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 98328 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P143524 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P143040 NM_146837.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P143040 NM_146837.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P143040 NM_146837.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P143040 NM_146837.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21405989 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21405982 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21405970 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21406009 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21405987 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21405980 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21405985 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21406015 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21405976 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21406012 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21405972 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21405991 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21406014 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21406013 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21406011 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21406010 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21405990 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21405968 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21405965 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21405978 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21405974 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21405998 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21405977 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21405994 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21406007 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21405986 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21405966 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21406004 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21405995 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21405963 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21406005 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P143524 - -
          Whole Mouse Genome G4122A A_51_P143040 NM_146837.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P143040 NM_146837.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P108918 NM_001011836.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P114906 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P143524 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01637182 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02218628 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01637184 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01637172 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02218634 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21405990 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21406004 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21406013 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21405995 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21406018 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21406014 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21405974 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21406038 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21406027 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21405982 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21406011 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21405966 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21405965 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21406009 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21405978 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21405968 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21405980 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21406031 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21406007 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21405986 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21406005 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21405987 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21405970 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21406033 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21405977 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21406012 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21406023 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21405972 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21405991 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21405985 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21406017 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21406024 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21405986 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01950463 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21406027 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21406023 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21406009 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21406034 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02218636 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21405976 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21406020 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21406012 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21405989 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21406013 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01765219 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02218628 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01147391 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21406028 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21405974 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21405987 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01368357 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21406031 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21405965 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21405980 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01368361 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21405995 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21405985 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21405990 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21405968 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01229795 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21406004 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21406016 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21406005 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21406011 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21405978 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02218623 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02218630 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21406014 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21406029 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21405972 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21406007 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21405991 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21405970 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21405982 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21405977 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10473554 - -
          Affymetrix Mouse Gene 1.0 ST Array 10473542 - -
          Affymetrix Mouse Gene 1.1 ST Array 10473553 - -
          Affymetrix Mouse Gene 1.1 ST Array 10473541 - -
          Affymetrix Mouse Clariom S Array PSR0200010041.mm.1 NM_146837.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200010041.mm.1 NM_146837.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200010041.mm.1 TC0200001293.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 472229 - -
          ABI Mouse Genome Survey Microarray 790042 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 3450086 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003450086 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2651036 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2651036 - -
          mouse-6 v1.1 (Illumina) scl0258834.1_29-S - -
          Illumina Sentrix mouse-6.v1 bead array 3450086 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0258834.1_29-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3450086 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1252123 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1252123 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline