Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00654475_s1
          See other OLFR1107 GE Assays ›
          Gene Symbol:
          Olfr1107
          Gene Name:
          olfactory receptor 1107
          Gene Aliases:
          MOR172-2
          Chromosome Location:
          Chr.2: 87071115 - 87072223 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00654475_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr1107
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146844.2 NP_667055.2 1 342 87
          GenBank mRNA BC120852.1 - 1 342 40129775 87
          BC137830.1 - 1 342 8860849 87

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200003969.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 G-protein coupled receptor Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr1107

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          G-protein coupled receptor activity
          olfactory receptor activity
          odorant binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2101992 NM_146844.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2101992 BC120852.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2101992 BC137830.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P105468 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2101992 NM_146844.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2101992 BC120852.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2101992 BC137830.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P182427 NM_146844.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P182427 BC120852.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P182427 BC137830.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P105468 - -
          Whole Mouse Genome G4122A A_51_P182427 NM_146844.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P182427 BC120852.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P182427 BC137830.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P104935 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P105468 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21404706 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21404701 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21404662 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21404697 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21404703 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21404689 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21404695 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21404696 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21404673 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21404711 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21404681 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21404651 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21404658 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21404687 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21404702 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21404691 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21404660 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21404680 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21404657 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21404676 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21404693 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21404685 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21404707 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21404678 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21404666 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21404686 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21404714 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21404663 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21404667 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21404650 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21404698 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21404693 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21404673 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21404658 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21404680 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21404676 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21404710 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21404714 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21404698 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21404706 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21404688 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21404681 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21404686 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21404687 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01508090 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21404707 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21404711 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02177225 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21404699 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21404665 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21404708 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21404689 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21404667 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01508066 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21404695 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21404678 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21404691 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21404701 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21404703 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21404660 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21404697 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21404662 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21404666 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21404657 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01508076 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21404685 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10484657 - -
          Affymetrix Mouse Gene 1.0 ST Array 10484656 - -
          Affymetrix Mouse Gene 1.0 ST Array 10484655 - -
          Affymetrix Mouse Gene 1.1 ST Array 10484654 - -
          Affymetrix Mouse Clariom S Array PSR0200033836.mm.1 NM_146844.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200033836.mm.1 NM_146844.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200033836.mm.1 TC0200003969.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200033837.mm.1 NM_146844.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200033837.mm.1 NM_146844.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200033837.mm.1 TC0200003969.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 616822 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 102940400 - -
          Illumina mouse-6 v1.0 expression beadchip 6420292 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006420292 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2685786 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2685786 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1256714 - -
          mouse-6 v1.1 (Illumina) scl4320.1.1_316-S - -
          mouse-6 v1.1 (Illumina) GI_22129178-S - -
          Illumina Sentrix mouse-6.v1 bead array 102940400 - -
          Illumina Sentrix mouse-6.v1 bead array 6420292 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl4320.1.1_316-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6420292 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 102940400 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2685786 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2685786 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-xpdfw:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline