Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00654510_s1
          See other OLFR898 GE Assays ›
          Gene Symbol:
          Olfr898
          Gene Name:
          olfactory receptor 898
          Gene Aliases:
          MOR170-3
          Chromosome Location:
          Chr.9: 38349085 - 38350035 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00654510_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr898
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146871.2 NP_667082.2 1 70 131
          GenBank mRNA BC127995.1 - 1 74 40135758 131

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr898

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          G-protein coupled receptor activity
          olfactory receptor activity
          odorant binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1959374 NM_146871.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1959374 BC127995.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1959374 NM_146871.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1959374 BC127995.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P371834 NM_146871.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P371834 BC127995.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26434431 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26434451 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26434430 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26434452 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26434426 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26434432 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26434428 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26434422 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26434425 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26434449 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26434455 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26434453 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26434424 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26434448 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26434435 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26434434 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26434438 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26434437 - -
          Whole Mouse Genome G4122A A_51_P371834 NM_146871.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P371834 BC127995.1 mouse_transcriptome
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07872690 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434435 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434466 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434452 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434432 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434430 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434457 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434425 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434449 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434426 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434455 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434431 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434461 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434419 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434470 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434472 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434434 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434459 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434428 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434467 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434453 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434437 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434423 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434424 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434462 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434457 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434452 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434449 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434464 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434467 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434428 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434438 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434435 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434437 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434455 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434470 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434453 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08110533 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434431 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434459 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434460 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434426 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07872690 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434469 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434447 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434448 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08062712 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434425 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434432 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434424 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434430 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434434 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434466 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434471 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434423 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10584400 - -
          Affymetrix Mouse Gene 1.1 ST Array 10584399 - -
          Affymetrix Mouse Clariom S Array PSR0900002791.mm.1 NM_146871.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900002791.mm.1 NM_146871.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0900002791.mm.1 TC0900000395.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 418067 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 6290487 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006290487 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1259210 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1259210 - -
          mouse-6 v1.1 (Illumina) scl37098.1.424_224-S - -
          Illumina Sentrix mouse-6.v1 bead array 6290487 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl37098.1.424_224-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6290487 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2742768 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2742768 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-jwmpg:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0