Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00654707_sH
          See other OLFR870 GE Assays ›
          Gene Symbol:
          Olfr870
          Gene Name:
          olfactory receptor 870
          Gene Aliases:
          MOR141-1, Olfr72
          Chromosome Location:
          Chr.9: 20170634 - 20171569 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00654707_sH
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr870
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146904.1 NP_667115.1 1 555 134

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr870

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P201751 NM_146904.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P201751 NM_146904.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1994579 BC119201.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1994579 BC119203.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1994579 NM_146903.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1994579 NM_146904.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1994579 BC119201.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1994579 BC119203.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1994579 NM_146903.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1994579 NM_146904.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P201751 NM_146904.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P201751 NM_146904.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P201751 NM_146904.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P201751 NM_146904.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26341846 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26341826 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26341849 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26341850 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26341856 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26341851 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26341859 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26341822 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26341848 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26341837 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26341845 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26341862 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26341855 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26341841 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26341824 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26341860 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26341828 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26341853 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26341838 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26341840 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26341843 - -
          Whole Mouse Genome G4122A A_51_P201751 NM_146904.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P201751 NM_146904.1 mouse_transcriptome
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07847518 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07981195 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07453150 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26341848 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26341860 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26341821 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26341851 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26341828 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26341859 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26341840 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26341866 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26341837 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26341864 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26341850 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26341819 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26341843 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26341841 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26341857 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26341863 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26341855 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26341824 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26341849 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26341853 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26341838 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26341845 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26341840 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26341851 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26341859 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26341841 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26341853 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26341860 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26341821 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26341857 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26341845 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26341856 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26341826 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26341839 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07905550 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26341824 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08181130 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26341837 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26341843 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07644755 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26341862 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26341855 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07981193 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26341819 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26341847 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26341848 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26341828 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10591217 - -
          Affymetrix HT MG-430 PM Array Plate 1450597_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10591216 - -
          Mouse Genome 430 2.0 Array 1450597_PM_at - -
          Mouse Genome 430 2.0 Array 1450597_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 57251_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 57251_at - -
          Mouse Expression Set 430 1450597_PM_at - -
          Mouse Expression Set 430 1450597_at - -
          Mouse Genome 430A 2.0 Array 1450597_PM_at - -
          Mouse Genome 430A 2.0 Array 1450597_at - -
          Affymetrix HT Mouse Genome 430A Array 1450597_at - -
          Affymetrix HT Mouse Genome 430A Array 1450597_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 887038 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 4780577 - -
          Illumina mouse-6 v1.0 expression beadchip 4780577 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004780577 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2726139 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2726139 - -
          mouse-6 v1.1 (Illumina) scl057251.1_6-S - -
          Illumina Sentrix mouse-6.v1 bead array 4780577 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl057251.1_6-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4780577 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2726139 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2726139 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline