Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00654773_s1
          See other OLFR346 GE Assays ›
          Gene Symbol:
          Olfr346
          Gene Name:
          olfactory receptor 346
          Gene Aliases:
          MOR136-11
          Chromosome Location:
          Chr.2: 36688004 - 36688933 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00654773_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr346
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146938.1 NP_667149.1 1 465 147

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr346

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception
          detection of chemical stimulus involved in sensory perception of smell
          transmembrane signaling receptor activity
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2069669 NM_146938.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2069669 NM_146938.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P503532 NM_146938.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21139750 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21139763 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21139749 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21139734 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21139741 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21139778 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21139773 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21139762 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21139775 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21139759 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21139746 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21139754 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21139783 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21139747 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21139740 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21139776 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21139782 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21139738 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21139737 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21139781 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21139743 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21139784 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21139752 - -
          Whole Mouse Genome G4122A A_51_P503532 NM_146938.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P132287 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01773597 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01773603 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01773580 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01773579 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01773578 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21139803 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21139746 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21139773 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21139756 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21139762 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21139792 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21139781 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21139797 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21139740 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21139786 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21139784 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21139782 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21139747 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21139741 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21139783 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21139745 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21139776 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21139737 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21139775 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21139759 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21139750 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21139789 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21139734 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21139741 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21139784 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21139756 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21139775 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21139762 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21139747 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21139780 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21139733 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21139759 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21139795 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21139773 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21139737 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21139743 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21139786 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21139776 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21139754 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21139740 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01287434 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01366419 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21139763 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21139781 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01160030 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01773591 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21139750 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21139752 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21139782 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21139746 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21139789 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21139790 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01254181 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21139783 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21139778 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10471752 - -
          Affymetrix Mouse Gene 1.1 ST Array 10471751 - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 838688 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 5570735 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005570735 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2599838 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2599838 - -
          mouse-6 v1.1 (Illumina) scl0258940.1_227-S - -
          Illumina Sentrix mouse-6.v1 bead array 5570735 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0258940.1_227-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5570735 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2599838 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2599838 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-mrft7:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0