Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00654829_s1
          See other OLFR340 GE Assays ›
          Gene Symbol:
          Olfr340
          Gene Name:
          olfactory receptor 340
          Gene Aliases:
          MOR136-1
          Chromosome Location:
          Chr.2: 36452587 - 36453525 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00654829_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr340
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146951.1 NP_667162.1 1 464 125
          GenBank mRNA BC109141.1 - 1 464 40041785 125
          BC109142.1 - 1 463 125

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr340

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception
          detection of chemical stimulus involved in sensory perception of smell
          transmembrane signaling receptor activity
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2046203 NM_146951.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2046203 BC109141.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2046203 BC109142.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P153193 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2046203 NM_146951.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2046203 BC109141.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2046203 BC109142.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P484859 NM_146951.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P484859 BC109141.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P484859 BC109142.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21139305 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21139318 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21139309 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21139337 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21139323 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21139319 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21139320 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21139317 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21139315 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21139321 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21139333 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21139343 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21139312 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21139308 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21139310 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21139341 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21139313 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21139311 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21139339 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P153193 - -
          Whole Mouse Genome G4122A A_51_P484859 NM_146951.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P484859 BC109141.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P484859 BC109142.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P102363 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P129118 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P153193 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01949438 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01949447 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01949444 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01949443 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21139347 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21139310 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21139355 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21139337 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21139345 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21139303 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21139319 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21139359 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21139339 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21139341 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21139335 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21139320 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21139315 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21139313 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21139311 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21139323 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21139343 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21139317 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21139308 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21139309 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21139305 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21139345 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21139351 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21139313 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21139310 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21139339 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21139335 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21139309 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21139343 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21139327 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21139359 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21139355 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21139356 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21139303 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21139332 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21139333 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21139320 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21139305 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21139317 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21139347 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21139323 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21139315 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21139352 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21139341 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21139357 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21139321 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21139349 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21139311 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21139312 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01949436 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01987403 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21139308 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21139337 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10471744 - -
          Affymetrix Mouse Gene 1.1 ST Array 10471743 - -
          Affymetrix Mouse Clariom S Array PSR0200006022.mm.1 NM_146951.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200006022.mm.1 NM_146951.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200006022.mm.1 TC0200000660.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 657402 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 3060524 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003060524 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2623440 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2623440 - -
          mouse-6 v1.1 (Illumina) scl5053.1.1_25-S - -
          Illumina Sentrix mouse-6.v1 bead array 3060524 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl5053.1.1_25-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3060524 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2623440 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2623440 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-jwmpg:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0