Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00654960_s1
          See other OLFR1262 GE Assays ›
          Gene Symbol:
          Olfr1262
          Gene Name:
          olfactory receptor 1262
          Gene Aliases:
          MOR234-1, or-leaf2
          Chromosome Location:
          Chr.2: 90002408 - 90003322 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00654960_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr1262
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146974.1 NP_667185.1 1 443 87
          GenBank mRNA AB074429.1 - 1 443 87
          BC120560.1 - 1 504 40129483 87
          BC120562.1 - 1 504 40129485 87

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 G-protein coupled receptor Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr1262

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception
          detection of chemical stimulus involved in sensory perception of smell
          transmembrane signaling receptor activity
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1953618 NM_146974.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1953618 AB074429.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1953618 BC120560.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1953618 BC120562.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P162985 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1953618 NM_146974.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1953618 AB074429.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1953618 BC120560.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1953618 BC120562.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P371411 NM_146974.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P371411 AB074429.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P371411 BC120560.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P371411 BC120562.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21415433 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21415449 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21415436 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21415453 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21415442 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21415440 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21415435 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21415418 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21415459 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21415467 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21415431 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21415447 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21415443 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21415464 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21415423 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21415444 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21415434 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21415438 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21415421 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21415441 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21415466 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21415437 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21415430 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21415420 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21415451 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P162985 - -
          Whole Mouse Genome G4122A A_51_P371411 NM_146974.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P371411 AB074429.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P371411 BC120560.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P371411 BC120562.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P131090 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P162985 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01437384 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01437387 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01437385 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415510 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415484 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415426 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415513 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415495 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415504 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415440 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415488 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415493 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415430 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415497 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415435 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415487 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415402 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415413 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415492 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415498 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415449 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415451 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415438 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415466 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415502 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415499 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415421 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415490 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415425 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415489 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415442 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415408 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415444 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415401 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415434 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415459 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415427 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415463 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415405 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415505 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415496 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415403 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415515 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415407 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415453 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415447 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415428 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415436 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415420 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415449 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01437390 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415420 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415433 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415429 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01437393 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415436 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415466 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415428 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415424 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415447 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415425 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415438 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415451 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415459 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415427 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415442 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01771684 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01437386 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415453 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415440 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415426 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01492834 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415419 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415467 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415430 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415421 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415423 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10473637 - -
          Affymetrix Mouse Gene 1.0 ST Array 10473638 - -
          Affymetrix Mouse Gene 1.1 ST Array 10473636 - -
          Affymetrix Mouse Clariom S Array PSR0200010158.mm.1 TC0200001341.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200010159.mm.1 NM_146974.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200010159.mm.1 NM_146974.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200010159.mm.1 TC0200001341.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 725296 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 2850441 - -
          Illumina mouse-6 v1.0 expression beadchip 2850441 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002850441 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2601330 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2601330 - -
          mouse-6 v1.1 (Illumina) scl20660.1.1_239-S - -
          Illumina Sentrix mouse-6.v1 bead array 2850441 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl20660.1.1_239-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2850441 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2601330 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2601330 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-6wvps:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0