Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00655069_s1
          See other OLFR394 GE Assays ›
          Gene Symbol:
          Olfr394
          Gene Name:
          olfactory receptor 394
          Gene Aliases:
          MOR135-8
          Chromosome Location:
          Chr.11: 73887438 - 73888370 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00655069_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr394
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_147007.1 NP_667218.1 1 472 150

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr394

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception
          detection of chemical stimulus involved in sensory perception of smell
          transmembrane signaling receptor activity
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1989149 NM_147007.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P161518 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1989149 NM_147007.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P328354 NM_147007.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27918353 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27918339 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27918357 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27918340 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27918354 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27918356 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27918365 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27918360 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27918363 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27918337 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27918342 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27918372 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27918335 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27918368 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27918341 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27918362 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27918366 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27918349 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27918343 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27918370 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27918364 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27918359 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P161518 - -
          Whole Mouse Genome G4122A A_51_P328354 NM_147007.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P124041 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P161518 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09469371 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09469365 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09469374 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27918363 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27918339 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27918366 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27918385 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27918359 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27918354 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27918382 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27918372 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27918365 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27918331 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27918349 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27918370 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27918380 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27918368 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27918357 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27918384 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27918335 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27918362 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27918337 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27918342 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27918364 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27918356 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27918360 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27918333 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27918341 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27918378 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27918384 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27918372 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27918339 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27918357 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27918359 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27918368 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27918382 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27918362 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27918337 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27918349 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27918333 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27918366 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09168419 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27918329 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27918353 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27918378 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09469348 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27918370 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27918365 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27918360 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27918341 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27918327 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27918377 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27918335 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27918356 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27918380 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27918364 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09469352 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27918331 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27918352 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27918336 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09168428 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27918385 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10388293 - -
          Affymetrix Mouse Gene 1.1 ST Array 10388292 - -
          Affymetrix Mouse Clariom S Array PSR1100029374.mm.1 NM_147007.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100029374.mm.1 NM_147007.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100029374.mm.1 TC1100003181.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 704701 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 4570022 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004570022 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1219111 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1219111 - -
          mouse-6 v1.1 (Illumina) scl13060.1.1_35-S - -
          Illumina Sentrix mouse-6.v1 bead array 4570022 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl13060.1.1_35-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4570022 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1219111 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1219111 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline