Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00655431_gH
          See other CYP4A12B GE Assays ›
          Gene Symbol:
          Cyp4a12b
          Gene Name:
          cytochrome P450, family 4, subfamily a, polypeptide 12B
          Gene Aliases:
          BC060945, Cyp4a12
          Chromosome Location:
          Chr.4: 115411624 - 115439034 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00655431_gH
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cyp4a12b
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_172306.2 NP_758510.2 6 - 7 839 109
          GenBank mRNA AK098123.1 - 6 - 7 820 109
          BC060945.1 - 6 - 7 820 30304458 109

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0400001223.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection P450 Detect Genomic DNA Recommended Assay Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cyp4a12b

          Panther Classification:

          Molecular Function -

          oxygenase

          Gene Ontology Categories:

          Function(s) Process(es)

          oxidation-reduction process
          monooxygenase activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2152607 AK098123.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2152607 NM_172306.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 86345 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 139939 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2152607 AK098123.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2152607 NM_172306.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23224322 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05366464 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P00964720 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P109939 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P134531 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P05366464 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23224416 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23224309 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03891836 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03516745 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03891837 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03934452 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23224315 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23224322 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23224326 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23224374 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23224321 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10507159 - -
          Affymetrix Mouse Gene 1.0 ST Array 10507157 - -
          Affymetrix Mouse Gene 1.0 ST Array 10507154 - -
          Affymetrix Mouse Gene 1.0 ST Array 10507158 - -
          Affymetrix Mouse Gene 1.0 ST Array 10507156 - -
          Affymetrix Mouse Gene 1.0 ST Array 10507153 - -
          Affymetrix Mouse Gene 1.0 ST Array 10507155 - -
          Affymetrix Mouse Gene 1.1 ST Array 10507152 - -
          Affymetrix Mouse Clariom S Array PSR0400008930.mm.1 NM_172306.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400008930.mm.1 NM_172306.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0400008930.mm.1 TC0400001223.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400008928.mm.1 NM_172306.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400008928.mm.1 NM_172306.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0400008928.mm.1 TC0400001223.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400008936.mm.1 NM_172306.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400008936.mm.1 NM_172306.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0400008936.mm.1 TC0400001223.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400008931.mm.1 NM_172306.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400008931.mm.1 NM_172306.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0400008931.mm.1 TC0400001223.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400008932.mm.1 NM_172306.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400008932.mm.1 NM_172306.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0400008932.mm.1 TC0400001223.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400008933.mm.1 NM_172306.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400008933.mm.1 NM_172306.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0400008933.mm.1 TC0400001223.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400008937.mm.1 NM_172306.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400008937.mm.1 NM_172306.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0400008937.mm.1 TC0400001223.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400008939.mm.1 NM_172306.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400008939.mm.1 NM_172306.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0400008939.mm.1 TC0400001223.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400008938.mm.1 NM_172306.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400008938.mm.1 NM_172306.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0400008938.mm.1 TC0400001223.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400008934.mm.1 NM_172306.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400008934.mm.1 NM_172306.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0400008934.mm.1 TC0400001223.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400008935.mm.1 NM_172306.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400008935.mm.1 NM_172306.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0400008935.mm.1 TC0400001223.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400008929.mm.1 NM_172306.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400008929.mm.1 NM_172306.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0400008929.mm.1 TC0400001223.mm.1 MTA TC
          Murine Genome U74v2 Set 101307_at - -
          Murine Genome U74v2 Set 101307_PM_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 6660170 - -
          Illumina mouse-6 v1.0 expression beadchip 6660170 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006660170 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1217072 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1217072 - -
          mouse-6 v1.1 (Illumina) scl013118.12_302-S - -
          Illumina Sentrix mouse-6.v1 bead array 6660170 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl013118.12_302-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6660170 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1217072 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1217072 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-7b6576f7bc-d59m7:80/100.66.79.221:80.
          git-commit: 6d309ebe7a31b2ceadeb87f43b473eebab110cf3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-2026.03.11-1.0