Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00655572_mH
          See other SLC22A29 GE Assays ›
          Gene Symbol:
          Slc22a29
          Gene Name:
          solute carrier family 22. member 29
          Gene Aliases:
          D630002G06Rik
          Chromosome Location:
          Chr.19: 8160168 - 8253519 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00655572_mH
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Slc22a29
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_172776.2 NP_766364.1 8 - 9 1561 150
          GenBank mRNA AF536196.1 - 9 - 10 1369 150
          AK052597.1 - 8 - 9 1562 150

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1900001767.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Transporter 3' Most Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Slc22a29

          Panther Classification:

          Molecular Function -

          secondary carrier transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P144531 AK052597.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P144531 NM_172776.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P144531 AF536195.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P144531 AF536196.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P144531 AK052597.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P144531 NM_172776.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P144531 AF536195.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P144531 AF536196.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 634807 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 840497 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 643433 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 891711 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 687360 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 133 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 76340 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 79154 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 136842 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P144531 AK052597.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P144531 NM_172776.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P144531 AF536195.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P144531 AF536196.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P144531 AK052597.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P144531 NM_172776.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P144531 AF536195.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P144531 AF536196.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31941473 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31941480 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31941475 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31941477 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31941472 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31941458 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09099542 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09099578 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09099523 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09099562 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09099567 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09099548 - -
          Whole Mouse Genome G4122A A_51_P144531 AK052597.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P144531 NM_172776.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P144531 AF536195.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P144531 AF536196.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P144531 AK052597.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P144531 NM_172776.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P144531 AF536195.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P144531 AF536196.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P125892 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P09099567 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P09099523 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P09099548 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P09099542 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32976455 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32976445 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32976447 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10465761 - -
          Affymetrix Mouse Gene 1.0 ST Array 10465756 - -
          Affymetrix Mouse Gene 1.0 ST Array 10465751 - -
          Affymetrix Mouse Gene 1.0 ST Array 10465748 - -
          Affymetrix Mouse Gene 1.0 ST Array 10465752 - -
          Affymetrix Mouse Gene 1.0 ST Array 10465759 - -
          Affymetrix Mouse Gene 1.0 ST Array 10465763 - -
          Affymetrix Mouse Gene 1.0 ST Array 10465760 - -
          Affymetrix Mouse Gene 1.0 ST Array 10465750 - -
          Affymetrix Mouse Gene 1.0 ST Array 10465757 - -
          Affymetrix Mouse Gene 1.0 ST Array 10465755 - -
          Affymetrix Mouse Gene 1.0 ST Array 10465762 - -
          Affymetrix Mouse Gene 1.0 ST Array 10465754 - -
          Affymetrix Mouse Gene 1.0 ST Array 10465744 - -
          Affymetrix Mouse Gene 1.0 ST Array 10465758 - -
          Affymetrix HT MG-430 PM Array Plate 1425222_PM_x_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10465753 - -
          Mouse Genome 430 2.0 Array 1425222_PM_x_at - -
          Mouse Genome 430 2.0 Array 1425222_x_at - -
          Mouse Genome 430 2.0 Array 1451635_PM_at - -
          Mouse Genome 430 2.0 Array 1451635_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 236293_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 236293_at - -
          Affymetrix Mouse Clariom S Array PSR1900009897.mm.1 NM_172776.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900009897.mm.1 NM_172776.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1900009897.mm.1 TC1900001767.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900009898.mm.1 NM_172776.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900009898.mm.1 NM_172776.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1900009898.mm.1 TC1900001767.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900009900.mm.1 NM_172776.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900009900.mm.1 NM_172776.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1900009900.mm.1 TC1900001767.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900009907.mm.1 NM_172776.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900009907.mm.1 NM_172776.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1900009907.mm.1 TC1900001767.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900009901.mm.1 NM_172776.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900009901.mm.1 NM_172776.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1900009901.mm.1 TC1900001767.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900009906.mm.1 NM_172776.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900009906.mm.1 NM_172776.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1900009906.mm.1 TC1900001767.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900009895.mm.1 NM_172776.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900009895.mm.1 NM_172776.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1900009895.mm.1 TC1900001767.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900009899.mm.1 NM_172776.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900009899.mm.1 NM_172776.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1900009899.mm.1 TC1900001767.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900009903.mm.1 NM_172776.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900009903.mm.1 NM_172776.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1900009903.mm.1 TC1900001767.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900009902.mm.1 NM_172776.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900009902.mm.1 NM_172776.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1900009902.mm.1 TC1900001767.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900009909.mm.1 NM_172776.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900009909.mm.1 NM_172776.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1900009909.mm.1 TC1900001767.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900009904.mm.1 NM_172776.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900009904.mm.1 NM_172776.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1900009904.mm.1 TC1900001767.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900009905.mm.1 NM_172776.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900009905.mm.1 NM_172776.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1900009905.mm.1 TC1900001767.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900009908.mm.1 NM_172776.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900009908.mm.1 NM_172776.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1900009908.mm.1 TC1900001767.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900009910.mm.1 NM_172776.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900009910.mm.1 NM_172776.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1900009910.mm.1 TC1900001767.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900009911.mm.1 NM_172776.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900009911.mm.1 NM_172776.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1900009911.mm.1 TC1900001767.mm.1 MTA TC
          Mouse Expression Set 430 1451635_PM_at - -
          Mouse Expression Set 430 1451635_at - -
          Mouse Genome 430A 2.0 Array 1451635_at - -
          Mouse Genome 430A 2.0 Array 1451635_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1425222_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1425222_PM_x_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 700251 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 4150026 - -
          Illumina mouse-6 v1.0 expression beadchip 4150026 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004150026 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1225143 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1225143 - -
          mouse-6 v1.1 (Illumina) scl00236293.1_126-S - -
          Illumina Sentrix mouse-6.v1 bead array 4150026 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl00236293.1_126-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4150026 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1225143 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline