Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00655735_m1
          See other CRYGD GE Assays ›
          Gene Symbol:
          Crygd
          Gene Name:
          crystallin, gamma D
          Gene Aliases:
          Aey4, Cryg-1, DGcry-1, Lop12
          Chromosome Location:
          Chr.1: 65061838 - 65063440 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00655735_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Crygd
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_007776.2 NP_031802.2 2 - 3 293 91
          GenBank mRNA AK136286.1 - 2 - 3 292 91
          BC057013.1 - 2 - 3 274 6824399 91
          BC132116.1 - 2 - 3 292 40130393 91
          BC132118.1 - 2 - 3 293 40130395 91
          CA452140.1 - 1 - 2 140 91
          K02583.1 - 2 - 3 262 91

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0100002477.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Crygd

          Gene Ontology Categories:

          Function(s) Process(es)

          eye development
          lens development in camera-type eye
          visual perception
          cellular response to reactive oxygen species
          lens fiber cell differentiation
          structural constituent of eye lens

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P483877 NM_007776.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P483877 AK136286.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P483877 BC057013.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P483877 BC132116.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P483877 BC132118.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P483877 NM_007776.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P483877 AK136286.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P483877 BC057013.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P483877 BC132116.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P483877 BC132118.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 305747 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 50120 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P125671 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P483877 NM_007776.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P483877 AK136286.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P483877 BC057013.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P483877 BC132116.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P483877 BC132118.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P483877 NM_007776.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P483877 AK136286.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P483877 BC057013.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P483877 BC132116.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P483877 BC132118.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P483877 NM_007776.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P483877 AK136286.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P483877 BC057013.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P483877 BC132116.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P483877 BC132118.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P483877 NM_007776.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P483877 AK136286.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P483877 BC057013.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P483877 BC132116.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P483877 BC132118.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20296226 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20296214 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20296212 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20296239 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20296218 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20296224 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20296199 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20296228 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20296292 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20296220 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20296217 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20296234 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20296194 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20296201 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20296225 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20296202 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20296210 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20296237 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20296209 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20296236 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20296233 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20296291 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20296216 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20296223 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20296230 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20296222 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20296294 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20296200 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20296207 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P125671 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P173430 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P00088078 - -
          Whole Mouse Genome G4122A A_51_P483877 NM_007776.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P483877 AK136286.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P483877 BC057013.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P483877 BC132116.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P483877 BC132118.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P483877 NM_007776.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P483877 AK136286.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P483877 BC057013.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P483877 BC132116.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P483877 BC132118.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P126655 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P173430 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P125671 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00551132 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00551163 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00551159 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00551144 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00551150 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20296237 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20296164 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20296200 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20296240 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20296216 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20296214 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20296199 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20296264 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20296233 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20296217 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20296194 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20296241 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20296228 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20296202 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20296220 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20296236 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20296201 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20296162 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20296258 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20296266 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20296231 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20296226 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20296229 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20296249 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20296154 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20296257 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20296167 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20296155 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20296209 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20296242 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20296218 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20296224 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20296255 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20296261 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20296212 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20296234 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20296243 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20296222 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20296225 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20296210 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20296248 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20296246 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20296254 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20296229 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20296257 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20296233 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20296212 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00800145 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20296216 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20296252 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00017256 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00867818 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20296225 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20296242 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20296202 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20296243 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20296210 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20296255 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20296220 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20296199 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20296231 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20296230 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20296193 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20296194 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20296241 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20296217 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20296223 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00800144 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20296224 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20296246 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20296226 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20296218 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20296222 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20296207 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20296201 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00551163 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20296214 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20296254 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20296240 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20296237 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20296200 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10355190 - -
          Affymetrix Mouse Gene 1.0 ST Array 10355192 - -
          Affymetrix HT MG-430 PM Array Plate 1419141_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10355191 - -
          Mouse Genome 430 2.0 Array 1419141_PM_at - -
          Mouse Genome 430 2.0 Array 1419141_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 12967_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 12967_at - -
          Affymetrix Mouse Clariom S Array PSR0100020114.mm.1 NM_007776.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100020114.mm.1 NM_007776.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0100020114.mm.1 TC0100002477.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100020118.mm.1 NM_007776.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100020118.mm.1 NM_007776.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0100020118.mm.1 TC0100002477.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100020117.mm.1 NM_007776.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100020117.mm.1 NM_007776.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0100020117.mm.1 TC0100002477.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100020113.mm.1 NM_007776.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100020113.mm.1 NM_007776.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0100020113.mm.1 TC0100002477.mm.1 MTA TC
          Mouse Expression Set 430 1419141_at - -
          Mouse Expression Set 430 1419141_PM_at - -
          Murine Genome U74v2 Set 96965_at - -
          Murine Genome U74v2 Set 104171_PM_f_at - -
          Murine Genome U74v2 Set 162268_at - -
          Murine Genome U74v2 Set 162018_PM_r_at - -
          Murine Genome U74v2 Set 96965_PM_at - -
          Murine Genome U74v2 Set 162268_PM_at - -
          Murine Genome U74v2 Set 162018_r_at - -
          Murine Genome U74v2 Set 104171_f_at - -
          Mouse Genome 430A 2.0 Array 1419141_PM_at - -
          Mouse Genome 430A 2.0 Array 1419141_at - -
          Affymetrix HT Mouse Genome 430A Array 1419141_at - -
          Affymetrix HT Mouse Genome 430A Array 1419141_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 438865 - -
          ABI Mouse Genome Survey Microarray 824287 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 2680095 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002680095 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2772482 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2772482 - -
          mouse-6 v1.1 (Illumina) scl012967.1_46-S - -
          Illumina Sentrix mouse-6.v1 bead array 2680095 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl012967.1_46-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2680095 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2884655 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2772482 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2884655 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline