Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00655745_m1
          See other CSF2RB GE Assays ›
          Gene Symbol:
          Csf2rb
          Gene Name:
          colony stimulating factor 2 receptor, beta, low-affinity (granulocyte-macrophage)
          Gene Aliases:
          AI848964, AIC2B, Bc, CDw131, Csf2rb1, Csfgmrb, Il3r, Il3rb, Il3rb1, Il5rb
          Chromosome Location:
          Chr.15: 78325797 - 78351001 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00655745_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Csf2rb
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_007780.4 NP_031806.3 3 - 4 471 160
          GenBank mRNA AF483496.1 - 2 - 3 213 160
          AF483497.1 - 2 - 3 213 160
          AK133420.1 - 415 160
          AK152608.1 - 3 - 4 411 160
          AK154286.1 - 3 - 4 408 160
          AK155109.1 - 3 - 4 442 160
          AK155215.1 - 3 - 4 405 160
          AK155699.1 - 3 - 4 408 160
          AK155803.1 - 3 - 4 408 160
          AK155822.1 - 3 - 4 411 160
          AK155885.1 - 3 - 4 412 160
          AK171477.1 - 3 - 4 408 160
          BC113202.1 - 4 - 5 482 40048156 160
          M34397.1 - 4 - 5 500 160

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1500000641.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Csf2rb

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          cytokine-mediated signaling pathway
          cytokine receptor activity
          protein binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1500004655.mm.1 TC1500000641.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500004662.mm.1 NM_007780.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500004662.mm.1 NM_007780.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1500004662.mm.1 TC1500000641.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500004664.mm.1 NM_007780.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500004664.mm.1 NM_007780.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1500004664.mm.1 TC1500000641.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500004660.mm.1 NM_007780.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500004660.mm.1 NM_007780.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1500004660.mm.1 TC1500000641.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500004659.mm.1 NM_007780.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500004659.mm.1 NM_007780.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1500004659.mm.1 TC1500000641.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500004661.mm.1 NM_007780.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500004661.mm.1 NM_007780.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1500004661.mm.1 TC1500000641.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500004663.mm.1 NM_007780.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500004663.mm.1 NM_007780.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1500004663.mm.1 TC1500000641.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500004657.mm.1 NM_007780.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500004657.mm.1 NM_007780.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1500004657.mm.1 TC1500000641.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 484140 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline