Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00655850_m1
          See other DEFA-RS1 GE Assays ›
          Gene Symbol:
          Defa-rs1
          Gene Name:
          defensin, alpha, related sequence 1
          Gene Aliases:
          2010300L12Rik, 2010319H24Rik, CRS1C, CRS1C-2, CRS1C-4, CRS1C-5, Defcr-rs1
          Chromosome Location:
          Chr.8: 21325888 - 21327020 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00655850_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Defa-rs1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_007844.2 NP_031870.2 1 - 2 213 109
          GenBank mRNA AK008584.1 - 1 - 2 214 109
          BC117042.1 - 1 - 2 188 40126361 109
          BC117044.1 - 1 - 2 188 40126363 109
          BC145556.1 - 1 - 2 218 9054120 109
          BC171966.1 - 1 - 2 218 9054252 109
          M33226.1 - 1 - 2 193 109

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Defa-rs1

          Panther Classification:

          Molecular Function -

          antimicrobial response protein

          Gene Ontology Categories:

          Function(s)

          defense response

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1979009 AV064248.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1979009 NM_001177482.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1979007 NM_007844.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1979007 AK008584.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1979007 NM_007844.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1979007 AK008584.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25706219 - -
          Whole Mouse Genome G4122A A_52_P592970 NM_001007582.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P592970 AK008498.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P592970 AY761184.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P592970 EU760895.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P178718 NM_001177526.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P178718 NM_001177518.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P117842 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P133967 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10577428 - -
          Affymetrix Mouse Gene 1.0 ST Array 10577422 - -
          Affymetrix Mouse Gene 1.0 ST Array 10577431 - -
          Affymetrix Mouse Gene 1.0 ST Array 10577435 - -
          Affymetrix Mouse Gene 1.0 ST Array 10577423 - -
          Affymetrix Mouse Gene 1.0 ST Array 10577436 - -
          Affymetrix Mouse Gene 1.0 ST Array 10577430 - -
          Affymetrix Mouse Gene 1.0 ST Array 10577427 - -
          Affymetrix HT MG-430 PM Array Plate 1418550_PM_x_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10577429 - -
          Affymetrix Mouse Gene 1.1 ST Array 10577434 - -
          Affymetrix Mouse Gene 1.1 ST Array 10577426 - -
          Affymetrix Mouse Gene 1.1 ST Array 10577421 - -
          Mouse Genome 430 2.0 Array 1418550_PM_x_at - -
          Mouse Genome 430 2.0 Array 1418550_x_at - -
          Affymetrix Mouse Clariom S Array PSR0800014590.mm.1 NM_007844.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800014590.mm.1 NM_007844.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0800014590.mm.1 TC0800001842.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800014591.mm.1 NM_007844.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800014591.mm.1 NM_007844.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0800014591.mm.1 TC0800001842.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800014588.mm.1 NM_007844.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800014588.mm.1 NM_007844.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0800014588.mm.1 TC0800001842.mm.1 MTA TC
          Mouse Expression Set 430 1418550_PM_x_at - -
          Mouse Expression Set 430 1418550_x_at - -
          Murine Genome U74v2 Set 92812_PM_f_at - -
          Murine Genome U74v2 Set 102814_PM_f_at - -
          Murine Genome U74v2 Set 92812_f_at - -
          Murine Genome U74v2 Set 102814_f_at - -
          Murine Genome U74v2 Set 167227_r_at - -
          Murine Genome U74v2 Set 165941_PM_at - -
          Murine Genome U74v2 Set 165941_at - -
          Murine Genome U74v2 Set 167227_PM_r_at - -
          Mouse Genome 430A 2.0 Array 1418550_x_at - -
          Mouse Genome 430A 2.0 Array 1418550_PM_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1418550_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1418550_PM_x_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 750672 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 107100142 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2524936 - -
          mouse-6 v1.1 (Illumina) GI_6681166-S - -
          Illumina Sentrix mouse-6.v1 bead array 107100142 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 107100142 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2993174 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2993174 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2524936 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-6wvps:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0