Search
Search

Vps4b| Gene Symbol: Vps4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009190.2 | NP_033216.2 | 6 - 7 | 916 | 149 | |
| GenBank mRNA | AK151023.1 | - | 6 - 7 | 870 | 149 | |
| AK152131.1 | - | 6 - 7 | 903 | 149 | ||
| AK165594.1 | - | 861 | 149 | |||
| AK168270.1 | - | 6 - 7 | 807 | 149 | ||
| BC003799.1 | - | 6 - 7 | 823 | 3488471 | 149 | |
| U10119.1 | - | 6 - 7 | 867 | 149 | ||
Set Membership: |
|