Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00656375_m1
          See other TNFRSF23 GE Assays ›
          Gene Symbol:
          Tnfrsf23
          Gene Name:
          tumor necrosis factor receptor superfamily, member 23
          Gene Aliases:
          SOB, TNFRSFH23, Tnfrh1, Tnfrsf1al1, mDcTRAILR1, mSOB
          Chromosome Location:
          Chr.7: 143665807 - 143686423 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00656375_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tnfrsf23
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AJ278264.1 - 3 - 4 286 77
          AK154786.1 - 3 - 4 312 77
          AY046550.1 - 3 - 4 295 77
          AY165625.1 - 3 - 4 301 77
          BC106828.1 - 3 - 4 297 40044453 77
          BC106829.1 - 3 - 4 301 40044455 77
          BG077775.2 - 3 - 4 287 77

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700004593.mm.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1974005 BG077775.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1974005 BY036522.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1974005 AK170037.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1974005 AY046550.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1974005 AY165625.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2071132 AK170037.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2071132 NM_024290.3 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 683848 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 189051 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 925070 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 856143 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 84959 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 59108 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P149961 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P172035 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1974005 BG077775.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1974005 BY036522.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1974005 AK170037.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1974005 AY046550.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1974005 AY165625.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2071132 AK170037.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2071132 NM_024290.3 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25602340 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25602342 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25602304 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25602320 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25602318 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25602337 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25602341 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25602338 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25602335 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25602305 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P172035 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P149961 - -
          Whole Mouse Genome G4122A A_51_P370720 BG077775.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P370720 AK170037.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P370720 AY165625.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P370720 BC106828.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P370720 BC106829.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P106748 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P172035 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P149961 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06368755 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06368753 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25602238 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25602335 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25602243 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25602251 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25602320 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25602245 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25602281 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25602236 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25602318 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25602232 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25602294 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25602280 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25602295 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25602342 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25602233 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25602341 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25602241 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25602337 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25602240 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25602317 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25602234 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25602304 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06490450 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25602340 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25602298 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25602334 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25602305 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25602335 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06368753 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25602295 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06490454 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25602293 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25602317 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25602320 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25602338 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25602337 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25602290 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25602294 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25602288 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25602302 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10569507 - -
          Affymetrix Mouse Gene 1.0 ST Array 10569511 - -
          Affymetrix Mouse Gene 1.0 ST Array 10569512 - -
          Affymetrix Mouse Gene 1.0 ST Array 10569506 - -
          Affymetrix Mouse Gene 1.0 ST Array 10569509 - -
          Affymetrix Mouse Gene 1.0 ST Array 10569505 - -
          Affymetrix Mouse Gene 1.0 ST Array 10569508 - -
          Affymetrix Mouse Gene 1.0 ST Array 10569510 - -
          Affymetrix HT MG-430 PM Array Plate 1422101_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10569504 - -
          Mouse Genome 430 2.0 Array 1422101_PM_at - -
          Mouse Genome 430 2.0 Array 1422101_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 79201_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 79201_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0700038843.mm.1 NM_024290.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0700038843.mm.1 TC0700004593.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700038837.mm.1 NM_024290.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0700038837.mm.1 TC0700004593.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700038838.mm.1 NM_024290.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0700038838.mm.1 TC0700004593.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700038841.mm.1 NM_024290.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0700038841.mm.1 TC0700004593.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700038839.mm.1 NM_024290.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0700038839.mm.1 TC0700004593.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700038842.mm.1 NM_024290.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0700038842.mm.1 TC0700004593.mm.1 MTA TC
          Mouse Expression Set 430 1422101_at - -
          Mouse Expression Set 430 1422101_PM_at - -
          Mouse Expression Set 430 1442590_at - -
          Mouse Expression Set 430 1442590_PM_at - -
          Mouse Genome 430A 2.0 Array 1422101_at - -
          Mouse Genome 430A 2.0 Array 1422101_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1422101_at - -
          Affymetrix HT Mouse Genome 430A Array 1422101_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 776650 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 2060408 - -
          Illumina mouse-6 v1.0 expression beadchip 2060408 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002060408 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1221454 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1221454 - -
          mouse-6 v1.1 (Illumina) scl30449.6.1_7-S - -
          Illumina Sentrix mouse-6.v1 bead array 2060408 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl30449.6.1_7-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2060408 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1221454 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1221454 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline