| Array Name |
Probe IDs |
Targets |
Target Type |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P211956 |
BC069842.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P211956 |
AF237915.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P211956 |
AK003110.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P211956 |
AK008968.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P211956 |
NM_025658.4 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P211956 |
BC069842.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P211956 |
AF237915.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P211956 |
AK003110.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P211956 |
AK008968.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P211956 |
NM_025658.4 |
mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
193447 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
887514 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
610345 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
556221 |
AK036470.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
556221 |
AK085836.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
556221 |
XM_483890.5 |
refseq_mrna_mouse_XM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
556221 |
XM_906658.3 |
refseq_mrna_mouse_XM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
26406 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
241927 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
309835 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
819015 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
345215 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
414842 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
23407 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
204507 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
96832 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
109172 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
158824 |
- |
- |
| Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P140426 |
- |
- |
| Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P149490 |
- |
- |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P211956 |
BC069842.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P211956 |
AF237915.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P211956 |
AK003110.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P211956 |
AK008968.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P211956 |
NM_025658.4 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P211956 |
BC069842.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P211956 |
AF237915.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P211956 |
AK003110.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P211956 |
AK008968.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P211956 |
NM_025658.4 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P294505 |
AK008968.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P294505 |
BC069842.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P294505 |
AF237915.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P294505 |
AK003110.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P294505 |
NM_025658.4 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P211956 |
BC069842.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P211956 |
AF237915.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P211956 |
AK003110.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P211956 |
AK008968.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P211956 |
NM_025658.4 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P211956 |
BC069842.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P211956 |
AF237915.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P211956 |
AK003110.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P211956 |
AK008968.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P211956 |
NM_025658.4 |
mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31956621 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31956611 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31956627 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31956628 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31956622 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31956632 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31956607 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31956599 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31956633 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31956634 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31956631 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31956615 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31956605 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31956606 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P140426 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P149490 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P09106655 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P09106683 |
- |
- |
| Whole Mouse Genome G4122A |
A_51_P294505 |
AK008968.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P294505 |
BC069842.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P294505 |
AF237915.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P294505 |
AK003110.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P294505 |
NM_025658.4 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P211956 |
BC069842.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P211956 |
AF237915.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P211956 |
AK003110.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P211956 |
AK008968.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P211956 |
NM_025658.4 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P211956 |
BC069842.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P211956 |
AF237915.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P211956 |
AK003110.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P211956 |
AK008968.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P211956 |
NM_025658.4 |
mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P130455 |
- |
- |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P140426 |
- |
- |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P149490 |
- |
- |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_67_P09106683 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P14392482 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P14392484 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31956734 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31956723 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31956662 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31956639 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31956750 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31956739 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31956682 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31956632 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31956683 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31956737 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31956753 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31956726 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31956685 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31956605 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31956599 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31956596 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31956618 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31956732 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31956650 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31956633 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31956631 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31956564 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31956688 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31956647 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31956645 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31956749 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31956703 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31956747 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31956736 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31956746 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31956607 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31956710 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31956730 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31956686 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31956627 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31956699 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31956562 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31956664 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31956700 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31956597 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31956563 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31956645 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31956595 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31956596 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31956618 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31956607 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P14280745 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31956739 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31956633 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31956603 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31956646 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31956736 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31956651 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31956734 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31956631 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31956632 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31956622 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31956647 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31956747 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31956627 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31956597 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P14393812 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31956611 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P14394884 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31956600 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P14429954 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31956635 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31956650 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31956605 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P14286115 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31956737 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P14429953 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P14359722 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31956741 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P14394883 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31956636 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31956746 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31956615 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31956643 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31956742 |
- |
- |