Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00656925_m1
          See other S100A9 GE Assays ›
          Gene Symbol:
          S100a9
          Gene Name:
          S100 calcium binding protein A9 (calgranulin B)
          Gene Aliases:
          60B8Ag, AW546964, BEE22, Cagb, GAGB, L1Ag, MRP14, p14
          Chromosome Location:
          Chr.3: 90692630 - 90695721 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00656925_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: S100a9
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001281852.1 NP_001268781.1 2 - 3 212 162
          NM_009114.3 NP_033140.1 2 - 3 209 162
          GenBank mRNA AK143826.1 - 2 - 3 183 162
          BC027635.1 - 1 - 2 161 1332797 162
          BY112406.1 - 2 - 3 179 162
          M83219.1 - 2 - 3 179 162

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0300002376.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene S100a9

          Panther Classification:

          Molecular Function -

          calmodulin-related

          Gene Ontology Categories:

          Function(s) Process(es)

          immune system process
          leukocyte migration involved in inflammatory response
          positive regulation of peptide secretion
          regulation of translation
          autophagy
          apoptotic process
          activation of cysteine-type endopeptidase activity involved in apoptotic process
          chemotaxis
          inflammatory response
          peptidyl-cysteine S-nitrosylation
          neutrophil chemotaxis
          leukocyte chemotaxis
          actin cytoskeleton reorganization
          peptidyl-cysteine S-trans-nitrosylation
          innate immune response
          regulation of integrin biosynthetic process
          regulation of inflammatory response
          positive regulation of inflammatory response
          neutrophil aggregation
          positive regulation of intrinsic apoptotic signaling pathway
          calcium ion binding
          microtubule binding
          zinc ion binding
          antioxidant activity
          Toll-like receptor 4 binding
          metal ion binding
          arachidonic acid binding
          RAGE receptor binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1998471 NM_009114.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1998471 AK143826.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1998471 BC027635.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1998471 M83219.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 26281 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 289840 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P108131 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P171671 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P122018 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1998471 NM_009114.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1998471 AK143826.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1998471 BC027635.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1998471 M83219.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P402943 NM_009114.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P402943 AK143826.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P402943 BC027635.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P402943 M83219.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22326624 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22326607 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22326615 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22326630 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22326617 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22326623 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22326609 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22326621 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22326606 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22326619 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22326599 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22326625 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22326602 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22326604 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22326605 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22326597 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22326603 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22326611 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22326628 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22326598 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22326632 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22326614 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P108131 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P122018 - -
          Whole Mouse Genome G4122A A_51_P402943 NM_009114.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P402943 AK143826.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P402943 BC027635.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P402943 M83219.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P126598 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P134748 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P108131 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P122018 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03087534 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03087540 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03087535 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03087536 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03087531 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03087542 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03087546 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03087543 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03087533 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22326628 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22326522 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22326634 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22326531 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22326581 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22326544 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22326560 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22326547 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22326585 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22326533 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22326606 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22326579 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22326520 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22326578 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22326590 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22326523 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22326599 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22326583 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22326614 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22326553 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22326521 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22326577 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22326630 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22326624 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22326525 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22326534 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22326556 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22326595 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22326562 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22326607 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22326623 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22326563 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22326559 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22326598 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22326594 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22326526 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22326565 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22326603 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22326619 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22326589 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22326575 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22326617 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22326588 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22326615 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22326529 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22326528 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22326605 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22326621 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22326611 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22326572 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22326524 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22326574 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22326536 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22326549 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22326541 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22326569 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22326573 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22326567 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22326638 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22326545 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22326636 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22326551 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22326593 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22326586 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22326538 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22326595 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22326593 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02619400 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03028656 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22326559 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22326636 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22326607 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03021637 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02388349 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22326579 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22326553 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02619388 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22326549 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22326585 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22326611 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22326621 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03087543 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22326618 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22326541 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22326572 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22326603 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03087535 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22326556 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22326598 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22326577 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22326569 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22326633 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02388361 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22326583 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22326606 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22326638 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22326578 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22326614 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02476480 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22326545 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22326631 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22326619 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02619377 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22326588 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22326589 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02619376 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22326623 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22326630 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22326544 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22326551 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22326634 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02619380 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02619383 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22326580 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22326605 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22326563 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22326615 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22326626 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22326567 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22326562 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02388353 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22326599 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03087530 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22326594 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02890293 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22326560 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22326565 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10499863 - -
          Affymetrix Mouse Gene 1.0 ST Array 10499862 - -
          Affymetrix HT MG-430 PM Array Plate 1448756_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10499861 - -
          Mouse Genome 430 2.0 Array 1448756_at - -
          Mouse Genome 430 2.0 Array 1448756_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 20202_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 20202_at - -
          Affymetrix Mouse Clariom S Array PSR0300018517.mm.1 NM_001281852.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300018517.mm.1 NM_009114.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0300018517.mm.1 TC0300002376.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300018518.mm.1 NM_001281852.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300018518.mm.1 NM_009114.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0300018518.mm.1 TC0300002376.mm.1 MTA TC
          Mouse Expression Set 430 1448756_at - -
          Mouse Expression Set 430 1448756_PM_at - -
          Murine Genome U74v2 Set 103887_at - -
          Murine Genome U74v2 Set 103887_PM_at - -
          Mouse Genome 430A 2.0 Array 1448756_at - -
          Mouse Genome 430A 2.0 Array 1448756_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1448756_at - -
          Affymetrix HT Mouse Genome 430A Array 1448756_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 811311 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 7050528 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0007050528 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2640519 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2640519 - -
          mouse-6 v1.1 (Illumina) scl020202.1_47-S - -
          Illumina Sentrix mouse-6.v1 bead array 7050528 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl020202.1_47-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 7050528 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2803674 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2803674 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl020202.1_47-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline