Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00656927_g1
          See other SAA1 GE Assays ›
          Gene Symbol:
          Saa1
          Gene Name:
          serum amyloid A 1
          Gene Aliases:
          Saa-1, Saa2
          Chromosome Location:
          Chr.7: 46740497 - 46743070 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00656927_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Saa1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_009117.3 NP_033143.1 3 - 4 262 147
          GenBank mRNA BC087933.1 - 3 - 4 262 30310171 147

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700003041.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Transporter 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Saa1

          Panther Classification:

          Molecular Function -

          apolipoprotein

          Gene Ontology Categories:

          Function(s) Process(es)

          acute-phase response
          cholesterol metabolic process
          positive chemotaxis
          cell chemotaxis
          G-protein coupled receptor binding
          protein binding
          heparin binding
          chemoattractant activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P318673 BC024606.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P318673 BC024606.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P318673 BC024606.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P318673 BC024606.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P361650 NM_009117.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P361650 AK136475.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P361650 BC087933.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25101051 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25101045 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25101063 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25101031 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25101029 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25101027 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25101050 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25101061 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25101049 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25101028 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25101064 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25101040 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25101036 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25101035 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25101048 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25101059 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25101072 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25101002 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25101065 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25101070 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25101010 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25101041 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25101062 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25101060 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25101056 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25101067 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25101030 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25101038 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25101043 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25101055 - -
          Whole Mouse Genome G4122A A_51_P361650 NM_009117.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P361650 AK136475.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P361650 BC087933.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P318673 BC024606.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P318673 BC024606.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P107235 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06382210 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06416448 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06025056 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05975080 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05924001 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06382206 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06382209 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25100972 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25101051 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25101060 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25101059 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25101049 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25101043 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25101072 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25101040 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25100948 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25101067 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25101063 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25100959 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25101027 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25101041 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25101030 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25100975 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25101056 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25101065 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25100951 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25100977 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25100968 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25100985 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25100965 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25100941 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25101068 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25101029 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25101038 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25100960 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25101073 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25100978 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25100944 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25100983 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25100980 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25100955 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25100942 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25101035 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25100961 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25101028 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25101045 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25100947 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25101048 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25101070 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25101055 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25100946 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25101045 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25101002 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25101049 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25101010 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06539700 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25101043 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25101028 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25101033 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06539702 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25101030 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25101013 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25101048 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25101036 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25101035 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25101050 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25101041 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06539717 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25101031 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06416451 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25101027 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06539696 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25101029 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10563614 - -
          Affymetrix Mouse Gene 1.0 ST Array 10563612 - -
          Affymetrix Mouse Gene 1.0 ST Array 10563613 - -
          Affymetrix HT MG-430 PM Array Plate 1450788_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1419075_PM_s_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10563611 - -
          Mouse Genome 430 2.0 Array 1419075_PM_s_at - -
          Mouse Genome 430 2.0 Array 1419075_s_at - -
          Mouse Genome 430 2.0 Array 1450788_at - -
          Mouse Genome 430 2.0 Array 1450788_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 20208_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 20208_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0700027764.mm.1 NM_009117.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700027764.mm.1 NM_009117.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0700027764.mm.1 TC0700003041.mm.1 MTA TC
          Mouse Expression Set 430 1450788_PM_at - -
          Mouse Expression Set 430 1450788_at - -
          Mouse Genome 430A 2.0 Array 1450788_PM_at - -
          Mouse Genome 430A 2.0 Array 1450788_at - -
          Affymetrix HT Mouse Genome 430A Array 1419075_PM_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1450788_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1419075_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1450788_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 5390520 - -
          Illumina mouse-6 v1.0 expression beadchip 5390520 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005390520 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2768154 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2768154 - -
          mouse-6 v1.1 (Illumina) scl020208.3_20-S - -
          Illumina Sentrix mouse-6.v1 bead array 5390520 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl020208.3_20-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5390520 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2826869 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2768154 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2826869 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl020208.3_20-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline