Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00657093_g1
          See other H2-Q1 GE Assays ›
          Gene Symbol:
          H2-Q1
          Gene Name:
          histocompatibility 2, Q region locus 1
          Gene Aliases:
          H-2Q1, Q1, Q1b, Q1d, Q1k, Qa-1, Qa1, Qed-1
          Chromosome Location:
          Chr.17: 35320516 - 35327699 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00657093_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: H2-Q1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_010390.3 NP_034520.2 2 - 3 334 124
          GenBank mRNA BC080812.1 - 2 - 3 345 6398887 124
          U96752.1 - 2 - 3 334 124

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene H2-Q1

          Panther Classification:

          Molecular Function -

          major histocompatibility complex protein

          Gene Ontology Categories:

          Function(s) Process(es)

          antigen processing and presentation of peptide antigen via MHC class I
          antigen processing and presentation
          receptor binding
          beta-2-microglobulin binding
          peptide binding
          peptide antigen binding
          T cell receptor binding
          poly(A) RNA binding
          TAP binding
          protein heterodimerization activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31158869 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31158873 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31158870 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31158874 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31158872 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1978502 U96752.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1978502 NM_010390.3 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P158147 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1978502 U96752.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1978502 NM_010390.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P231729 AK080269.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P231729 U96752.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P231729 NM_010390.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P231729 BC028930.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P231729 BC080812.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P425048 BC028930.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P425048 BC080812.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P425048 BC028930.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P425048 BC080812.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31158863 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31158875 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31158874 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31158885 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31158870 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31158873 - -
          Whole Mouse Genome G4122A A_52_P231729 AK080269.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P231729 U96752.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P231729 NM_010390.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P231729 BC028930.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P231729 BC080812.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P425048 BC028930.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P425048 BC080812.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P425048 BC028930.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P425048 BC080812.1 mouse_transcriptome
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13256436 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31158870 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31158873 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31158872 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31158869 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31158874 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31158931 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31158938 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31158937 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31158869 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31158926 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31158873 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31158884 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31158870 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31158874 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31158854 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31158848 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31158853 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31158849 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31158935 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31158872 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31158878 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31158875 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31158884 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31158891 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31158893 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31158874 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13256428 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31158873 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31158872 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31158878 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13256436 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13077000 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31158885 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31158863 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10444796 - -
          Affymetrix Mouse Gene 1.0 ST Array 10444794 - -
          Affymetrix Mouse Gene 1.0 ST Array 10444790 - -
          Affymetrix Mouse Gene 1.0 ST Array 10444792 - -
          Affymetrix Mouse Gene 1.0 ST Array 10444795 - -
          Affymetrix Mouse Gene 1.0 ST Array 10444798 - -
          Affymetrix Mouse Gene 1.0 ST Array 10444791 - -
          Affymetrix Mouse Gene 1.0 ST Array 10444793 - -
          Affymetrix HT MG-430 PM Array Plate 1418734_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10444788 - -
          Mouse Genome 430 2.0 Array 1418734_at - -
          Mouse Genome 430 2.0 Array 1451593_PM_at - -
          Mouse Genome 430 2.0 Array 1451644_a_at - -
          Mouse Genome 430 2.0 Array 1451644_PM_a_at - -
          Mouse Genome 430 2.0 Array 1418734_PM_at - -
          Mouse Genome 430 2.0 Array 1451593_at - -
          Affymetrix Mouse Clariom S Array PSR1700006996.mm.1 NM_010390.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700006996.mm.1 NM_010390.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1700006996.mm.1 TC1700002791.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700006997.mm.1 NM_010390.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700006997.mm.1 NM_010390.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1700006997.mm.1 TC1700002791.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700024758.mm.1 NM_010390.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700024758.mm.1 NM_010390.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1700024758.mm.1 TC1700002791.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700007005.mm.1 NM_010390.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700007005.mm.1 NM_010390.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1700007005.mm.1 TC1700002791.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700006994.mm.1 NM_010390.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700006994.mm.1 NM_010390.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1700006994.mm.1 TC1700002791.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700007003.mm.1 NM_010390.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700007003.mm.1 NM_010390.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1700007003.mm.1 TC1700002791.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700006993.mm.1 NM_010390.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700006993.mm.1 NM_010390.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1700006993.mm.1 TC1700002791.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700007001.mm.1 NM_010390.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700007001.mm.1 NM_010390.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1700007001.mm.1 TC1700002791.mm.1 MTA TC
          Mouse Expression Set 430 1431008_at - -
          Mouse Expression Set 430 1431008_PM_at - -
          Mouse Expression Set 430 1451644_a_at - -
          Mouse Expression Set 430 1451644_PM_a_at - -
          Mouse Expression Set 430 1418734_PM_at - -
          Mouse Expression Set 430 1418734_at - -
          Murine Genome U74v2 Set 99378_f_at - -
          Murine Genome U74v2 Set 97173_PM_f_at - -
          Murine Genome U74v2 Set 92222_PM_f_at - -
          Murine Genome U74v2 Set 97173_f_at - -
          Murine Genome U74v2 Set 92222_f_at - -
          Murine Genome U74v2 Set 99378_PM_f_at - -
          Mouse Genome 430A 2.0 Array 1451644_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1418734_PM_at - -
          Mouse Genome 430A 2.0 Array 1451644_a_at - -
          Mouse Genome 430A 2.0 Array 1418734_at - -
          Mouse Genome 430A 2.0 Array 1431008_PM_at - -
          Mouse Genome 430A 2.0 Array 1431008_at - -
          Affymetrix HT Mouse Genome 430A Array 1418734_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1418734_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 104810333 - -
          Illumina mouse-6 v1.0 expression beadchip 104810333 - -
          Illumina mouse-6 v1.0 expression beadchip 2470112 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002470112 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1216688 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1241491 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1216688 - -
          mouse-6 v1.1 (Illumina) GI_6754133-S - -
          mouse-6 v1.1 (Illumina) scl0015006.1_287-S - -
          Illumina Sentrix mouse-6.v1 bead array 2470112 - -
          Illumina Sentrix mouse-6.v1 bead array 104810333 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0015006.1_287-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 104810333 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2470112 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1216688 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1216688 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1241491 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline